Merck
  • Spatial control of nucleoporin condensation by fragile X-related proteins.
Spatial control of nucleoporin condensation by fragile X-related proteins.
The EMBO journal (2020-07-25)
Arantxa Agote-Aran, Stephane Schmucker, Katerina Jerabkova, Inès Jmel Boyer, Alessandro Berto, Laura Pacini, Paolo Ronchi, Charlotte Kleiss, Laurent Guerard, Yannick Schwab, Hervé Moine, Jean-Louis Mandel, Sebastien Jacquemont, Claudia Bagni, Izabela Sumara
ABSTRACT
Nucleoporins (Nups) build highly organized nuclear pore complexes (NPCs) at the nuclear envelope (NE). Several Nups assemble into a sieve-like hydrogel within the central channel of the NPCs. In the cytoplasm, the soluble Nups exist, but how their assembly is restricted to the NE is currently unknown. Here, we show that fragile X-related protein 1 (FXR1) can interact with several Nups and facilitate their localization to the NE during interphase through a microtubule-dependent mechanism. Downregulation of FXR1 or closely related orthologs FXR2 and fragile X mental retardation protein (FMRP) leads to the accumulation of cytoplasmic Nup condensates. Likewise, models of fragile X syndrome (FXS), characterized by a loss of FMRP, accumulate Nup granules. The Nup granule-containing cells show defects in protein export, nuclear morphology and cell cycle progression. Our results reveal an unexpected role for the FXR protein family in the spatial regulation of nucleoporin condensation.
Sigma-Aldrich
Anti-GAPDH antibody produced in rabbit, ~1 mg/mL, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Anti-Lamin A (C-terminal) antibody produced in rabbit, ~1 mg/mL, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Monastrol, ≥98% (HPLC), solid
Sigma-Aldrich
RIPAb+ FXR1 - RIP Validated Antibody and Primer Set, from mouse
Sigma-Aldrich
Collagen Type I, rat tail
Sigma-Aldrich
1,6-Hexanediol solution, BioUltra, ~6 M in H2O
Sigma-Aldrich
Anti-Dynein Antibody, 74 kDa Intermediate chains, cytoplasmic, clone 74.1, clone 74.1, Chemicon®, from mouse
Sigma-Aldrich
Anti-BICD2 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution, Ab1
Sigma-Aldrich
Anti-FXR1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution