Merck
  • A GDSL lipase-like from Ipomoea batatas catalyzes efficient production of 3,5-diCQA when expressed in Pichia pastoris.
A GDSL lipase-like from Ipomoea batatas catalyzes efficient production of 3,5-diCQA when expressed in Pichia pastoris.
Communications biology (2020-11-15)
Sissi Miguel, Guillaume Legrand, Léonor Duriot, Marianne Delporte, Barbara Menin, Cindy Michel, Alexandre Olry, Gabrielle Chataigné, Aleksander Salwinski, Joakim Bygdell, Dominique Vercaigne, Gunnar Wingsle, Jean Louis Hilbert, Frédéric Bourgaud, Alain Hehn, David Gagneul
ABSTRACT
The synthesis of 3,5-dicaffeoylquinic acid (3,5-DiCQA) has attracted the interest of many researchers for more than 30 years. Recently, enzymes belonging to the BAHD acyltransferase family were shown to mediate its synthesis, albeit with notably low efficiency. In this study, a new enzyme belonging to the GDSL lipase-like family was identified and proven to be able to transform chlorogenic acid (5-O-caffeoylquinic acid, 5-CQA, CGA) in 3,5-DiCQA with a conversion rate of more than 60%. The enzyme has been produced in different expression systems but has only been shown to be active when transiently synthesized in Nicotiana benthamiana or stably expressed in Pichia pastoris. The synthesis of the molecule could be performed in vitro but also by a bioconversion approach beginning from pure 5-CQA or from green coffee bean extract, thereby paving the road for producing it on an industrial scale.