Merck
  • Involvement of NDPK-B in Glucose Metabolism-Mediated Endothelial Damage via Activation of the Hexosamine Biosynthesis Pathway and Suppression of O-GlcNAcase Activity.
Involvement of NDPK-B in Glucose Metabolism-Mediated Endothelial Damage via Activation of the Hexosamine Biosynthesis Pathway and Suppression of O-GlcNAcase Activity.
Cells (2020-10-23)
Anupriya Chatterjee, Rachana Eshwaran, Gernot Poschet, Santosh Lomada, Mahmoud Halawa, Kerstin Wilhelm, Martina Schmidt, Hans-Peter Hammes, Thomas Wieland, Yuxi Feng
ABSTRACT
Our previous studies identified that retinal endothelial damage caused by hyperglycemia or nucleoside diphosphate kinase-B (NDPK-B) deficiency is linked to elevation of angiopoietin-2 (Ang-2) and the activation of the hexosamine biosynthesis pathway (HBP). Herein, we investigated how NDPK-B is involved in the HBP in endothelial cells (ECs). The activities of NDPK-B and O-GlcNAcase (OGA) were measured by in vitro assays. Nucleotide metabolism and O-GlcNAcylated proteins were assessed by UPLC-PDA (Ultra-performance liquid chromatography with Photodiode array detection) and immunoblot, respectively. Re-expression of NDPK-B was achieved with recombinant adenoviruses. Our results show that NDPK-B depletion in ECs elevated UDP-GlcNAc levels and reduced NDPK activity, similar to high glucose (HG) treatment. Moreover, the expression and phosphorylation of glutamine:fructose-6-phosphate amidotransferase (GFAT) were induced, whereas OGA activity was suppressed. Furthermore, overall protein O-GlcNAcylation, along with O-GlcNAcylated Ang-2, was increased in NDPK-B depleted ECs. Pharmacological elevation of protein O-GlcNAcylation using Thiamet G (TMG) or OGA siRNA increased Ang-2 levels. However, the nucleoside triphosphate to diphosphate (NTP/NDP) transphosphorylase and histidine kinase activity of NDPK-B were dispensable for protein O-GlcNAcylation. NDPK-B deficiency hence results in the activation of HBP and the suppression of OGA activity, leading to increased protein O-GlcNAcylation and further upregulation of Ang-2. The data indicate a critical role of NDPK-B in endothelial damage via the modulation of the HBP.
Sigma-Aldrich
MISSION® esiRNA, targeting human MGEA5
Millipore
Gelatin from porcine skin, suitable for microbiology, low gel strength
Sigma-Aldrich
Thiamet G, ≥98% (HPLC)
Sigma-Aldrich
Anti-Mouse IgG (whole molecule)–Peroxidase antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
Sigma-Aldrich
Puromycin dihydrochloride from Streptomyces alboniger, ≥98% (HPLC), powder
Sigma-Aldrich
Anti-γ-Tubulin antibody, Mouse monoclonal, clone GTU-88, ascites fluid
Sigma-Aldrich
Penicillin-Streptomycin, Solution stabilized, with 10,000 units penicillin and 10 mg streptomycin/mL, 0.1 μm filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Anti-N1-Phosphohistidine (1-pHis) Antibody, clone SC1-1, clone SC1-1, from rabbit