Merck
  • Functional Attributes and Anticancer Potentialities of Chico (Pachycereus Weberi) and Jiotilla (Escontria Chiotilla) Fruits Extract.
Functional Attributes and Anticancer Potentialities of Chico (Pachycereus Weberi) and Jiotilla (Escontria Chiotilla) Fruits Extract.
Plants (Basel, Switzerland) (2020-12-04)
Luisaldo Sandate-Flores, Eduardo Romero-Esquivel, José Rodríguez-Rodríguez, Magdalena Rostro-Alanis, Elda M Melchor-Martínez, Carlos Castillo-Zacarías, Patricia Reyna Ontiveros, Marcos Fredy Morales Celaya, Wei-Ning Chen, Hafiz M N Iqbal, Roberto Parra-Saldívar
ABSTRACT
Mexico has a great diversity of cacti, however, many of their fruits have not been studied in greater depth. Several bioactive compounds available in cacti juices extract have demonstrated nutraceutical properties. Two cactus species are interesting for their biologically active pigments, which are chico (Pachycereus weberi (J. M. Coult.) Backeb)) and jiotilla (Escontria chiotilla (Weber) Rose)). Hence, the goal of this work was to evaluate the bioactive compounds, i.e., betalains, total phenolic, vitamin C, antioxidant, and mineral content in the extract of the above-mentioned P. weberi and E. chiotilla. Then, clarified extracts were evaluated for their antioxidant activity and cytotoxicity (cancer cell lines) potentialities. Based on the obtained results, Chico fruit extract was found to be a good source of vitamin C (27.19 ± 1.95 mg L-Ascorbic acid/100 g fresh sample). Moreover, chico extract resulted in a high concentration of micronutrients, i.e., potassium (517.75 ± 16.78 mg/100 g) and zinc (2.46 ± 0.65 mg/100 g). On the other hand, Jiotilla has a high content of biologically active pigment, i.e., betaxanthins (4.17 ± 0.35 mg/g dry sample). The antioxidant activities of clarified extracts of chico and jiotilla were 80.01 ± 5.10 and 280.88 ± 7.62 mg/100 g fresh sample (DPPH method), respectively. From the cytotoxicity perspective against cancer cell lines, i.e., CaCo-2, MCF-7, HepG2, and PC-3, the clarified extracts of chico showed cytotoxicity (%cell viability) in CaCo-2 (49.7 ± 0.01%) and MCF-7 (45.56 ± 0.05%). A normal fibroblast cell line (NIH/3T3) was used, as a control, for comparison purposes. While jiotilla extract had cytotoxicity against HepG2 (47.31 ± 0.03%) and PC-3 (53.65 ± 0.04%). These results demonstrated that Chico and jiotilla are excellent resources of biologically active constituents with nutraceuticals potentialities.
Sigma-Aldrich
2,2′-Azino-bis(3-ethylbenzothiazoline-6-sulfonic acid) diammonium salt, tablet, 10 mg substrate per tablet
Sigma-Aldrich
Folin & Ciocalteu′s phenol reagent, suitable for determination of total protein by Lowry method, 1.9-2.1 N
Sigma-Aldrich
2,2-Diphenyl-1-picrylhydrazyl
Sigma-Aldrich
Caffeic acid, ≥98.0% (HPLC)
Sigma-Aldrich
Fumaric acid, ≥99.0% (T)
Sigma-Aldrich
Potassium persulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Potassium phosphate monobasic, ACS reagent, ≥99.0%
Sigma-Aldrich
Gallic acid monohydrate, ACS reagent, ≥98.0%
Supelco
2,4,6-Tris(2-pyridyl)-s-triazine, for spectrophotometric det. (of Fe), ≥98%
Sigma-Aldrich
p-Coumaric acid, ≥98.0% (HPLC)
Sigma-Aldrich
Sodium carbonate, BioXtra, ≥99.0%
Sigma-Aldrich
Iron(III) chloride hexahydrate, reagent grade, ≥98%, chunks