Merck
  • Cryptococcus gattii alters immunostimulatory potential in response to the environment.
Cryptococcus gattii alters immunostimulatory potential in response to the environment.
PloS one (2019-08-10)
Keigo Ueno, Yoshiko Otani, Nao Yanagihara, Takumi Nakamura, Kiminori Shimizu, Satoshi Yamagoe, Yoshitsugu Miyazaki
ABSTRACT
Cryptococcus gattii is a capsular fungal pathogen, which causes life-threatening cryptococcosis in immunocompetent individuals. This emerging pathogen is less likely to be recognized by innate immunity compared to traditional Cryptococcus neoformans strains. Previous studies indicate that C-type lectin receptors (CLRs), including dectin-1 and dectin-2, play a role in recognizing cryptococcal cells; however, it remains to be elucidated whether the receptors physically associate with C. gattii yeast cell surfaces. Based on the previous findings, we hypothesized that culture conditions influence the expression or exposure of CLR ligands on C. gattii. Therefore, in the present study, we first investigated the culture conditions that induce exposure of CLR ligands on C. gattii yeast cells via the binding assay using recombinant fusion proteins of mouse CLR and IgG Fc, Fc dectin-1 and Fc dectin-2. Common fungal culture media, such as yeast extract-peptone-dextrose (YPD) broth, Sabouraud broth, and potato dextrose agar, did not induce the exposure of dectin-1 ligands, including β-1,3-glucan, on both capsular and acapsular C. gattii strains, in contrast to Fc dectin-1 and Fc dectin-2 bound to C. gattii cells growing in the conventional synthetic dextrose (SD) medium [may also be referred to as a yeast nitrogen base with glucose medium]. The medium also induced the exposure of dectin-1 ligands on C. neoformans, whereas all tested media induced dectin-1 and dectin-2 ligands in a control fungus Candida albicans. Notably, C. gattii did not expose dectin-1 ligands in SD medium supplemented with yeast extract or neutral buffer. In addition, compared to YPD medium-induced C. gattii, SD medium-induced C. gattii more efficiently induced the phosphorylation of Syk, Akt, and Erk1/2 in murine dendritic cells (DCs). Afterwards, the cells were considerably engulfed by DCs and remarkably induced DCs to secrete the inflammatory cytokines. Overall, the findings suggest that C. gattii alters its immunostimulatory potential in response to the environment.
Sigma-Aldrich
Anti-Glucuronoxylomannan (GXM) Antibody, clone 18B7, clone 18B7, from mouse