Merck
Bioactive guaianolides from siyekucai (Ixeris chinensis).
Journal of natural products (2006-10-28)
Shujun Zhang, Ming Zhao, Liming Bai, Toshiaki Hasegawa, Jinlan Wang, Liyan Wang, Hongyan Xue, Qigang Deng, Fenglan Xing, Yuhua Bai, Jun-ichi Sakai, Jiao Bai, Rei Koyanagi, Yoshinori Tsukumo, Takao Kataoka, Kazuo Nagai, Katutoshi Hirose, Masayoshi Ando
ABSTRACT
Two new guaianolides, named chinensiolides D (5) and E (6), were isolated from Ixeris chinensis Nakai, and their structures were determined to be 10alpha-hydroxy-3-oxoguaia-11(13)-eno-12,6alpha-lactone (5) and 10alpha-hydroxy-3beta-O-[2,6-di(p-hydroxyphenylacetyl)-beta-glucopylanosyl]guaia-4(15),11(13)-dieno-12,6alpha-lactone (6). The first isolation of (11S)-10alpha-hydroxy-3-oxoguaia-4-eno-12,6alpha-lactone (4) from natural sources and its characterization are also reported. Chinenciolide E (6) showed significant growth inhibitory activity toward VA-13 malignant lung tumor cells (IC50 = 0.72 microM).
Sigma-Aldrich
Doxorubicin hydrochloride, suitable for fluorescence, 98.0-102.0% (HPLC)
Sigma-Aldrich
Doxorubicin hydrochloride, 98.0-102.0% (HPLC)