Merck
  • Carbonic anhydrase activators: activation of isozyme XIII with amino acids and amines.
Carbonic anhydrase activators: activation of isozyme XIII with amino acids and amines.
Bioorganic & medicinal chemistry letters (2006-05-30)
Seppo Parkkila, Daniela Vullo, Luca Puccetti, Anna-Kaisa Parkkila, Andrea Scozzafava, Claudiu T Supuran
ABSTRACT
The first activation study of isoform XIII of carbonic anhydrase (CA, EC 4.2.1.1) is reported. A series of amino acids and amines incorporating protonatable moieties of the primary/heterocyclic amine type were included in the study. As for CA I and II, CA XIII activators enhance kcat and show no effect on KM, for the physiologic reaction catalyzed by this isoform. Excellent CA XIII activating properties were shown by D-amino acids (His, Phe, DOPA, and Trp), serotonin, and 4-(2-aminoethyl)-morpholine, whereas the corresponding L-amino acids, dopamine, histamine, and 1-(2-aminoethyl)-piperazine, were weaker activators.
Product Description
Supelco
L-Histidine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
L-Histidine, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
1-(2-Aminoethyl)piperazine, 99%
SAFC
L-Histidine
Sigma-Aldrich
D-Histidine, ≥98% (TLC)
Sigma-Aldrich
Histamine, ≥97.0%
Sigma-Aldrich
D-Tryptophan, ≥98.0% (HPLC)
Sigma-Aldrich
D-Phenylalanine, ≥98% (HPLC)
Sigma-Aldrich
L-Histidine, suitable for cell culture, meets EP, USP testing specifications, from non-animal source
Sigma-Aldrich
L-Histidine, ReagentPlus®, ≥99% (TLC)
Sigma-Aldrich
4-(2-Aminoethyl)morpholine, 99%
Sigma-Aldrich
2-(2-Pyridyl)ethylamine, 95%
Sigma-Aldrich
L-Tryptophan, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
L-Phenylalanine, BioUltra, ≥99.0% (NT)
Sigma-Aldrich
L-Phenylalanine, 99%, FCC
Sigma-Aldrich
2-Picolylamine, 99%
Sigma-Aldrich
3,4-Dihydroxy-D-phenylalanine, powder, ≥95%
Sigma-Aldrich
L-Phenylalanine, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 98.5-101.0%
Sigma-Aldrich
L-Phenylalanine, reagent grade, ≥98%
Sigma-Aldrich
L-Tryptophan, reagent grade, ≥98% (HPLC)
Sigma-Aldrich
L-Tryptophan, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 99.0-101.0%
Sigma-Aldrich
3,4-Dihydroxy-L-phenylalanine, ≥98% (TLC)
Supelco
L-Phenylalanine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
L-Tryptophan, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland