Merck
  • Single-cell lineage analysis reveals extensive multimodal transcriptional control during directed beta-cell differentiation.
Single-cell lineage analysis reveals extensive multimodal transcriptional control during directed beta-cell differentiation.
Nature metabolism (2020-12-02)
Chen Weng, Jiajia Xi, Haiyan Li, Jian Cui, Anniya Gu, Sisi Lai, Konstantin Leskov, Luxin Ke, Fulai Jin, Yan Li, Chen Weng, Jiajia Xi, Haiyan Li, Jian Cui, Anniya Gu, Sisi Lai, Konstantin Leskov, Luxin Ke, Fulai Jin, Yan Li
ABSTRACT
The in vitro differentiation of insulin-producing beta-like cells can model aspects of human pancreatic development. Here, we generate 95,308 single-cell transcriptomes and reconstruct a lineage tree of the entire differentiation process from human embryonic stem cells to beta-like cells to study temporally regulated genes during differentiation. We identify so-called 'switch genes' at the branch point of endocrine/non-endocrine cell fate choice, revealing insights into the mechanisms of differentiation-promoting reagents, such as NOTCH and ROCKII inhibitors, and providing improved differentiation protocols. Over 20% of all detectable genes are activated multiple times during differentiation, even though their enhancer activation is usually unimodal, indicating extensive gene reuse driven by different enhancers. We also identify a stage-specific enhancer at the TCF7L2 locus for diabetes, uncovered by genome-wide association studies, that drives a transient wave of gene expression in pancreatic progenitors. Finally, we develop a web app to visualize gene expression on the lineage tree, providing a comprehensive single-cell data resource for researchers studying islet biology and diabetes.
Sigma-Aldrich
SANT-1, ≥98% (HPLC), powder
Sigma-Aldrich
Retinoic acid, ≥98% (HPLC), powder
Sigma-Aldrich
Zinc sulfate heptahydrate, BioReagent, suitable for cell culture
Liothyronine sodium, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
N-Acetyl-L-cysteine, BioReagent, suitable for cell culture
Sigma-Aldrich
3,3′,5-Triiodo-L-thyronine sodium salt, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
γ-Secretase Inhibitor XX, This γ-secretase inhibitor, CAS 209984-56-5, is a cell-permeable dibenzazepine compound that lowers both brain and plasma Aβ40 levels by ~72% in Tg2576 mutant APP transgenic mouse model.
Sigma-Aldrich
Heparin sodium salt from porcine intestinal mucosa, Grade I-A, ≥180 USP units/mg, powder, BioReagent, suitable for cell culture