Merck
  • Distinct roles of ezrin, radixin and moesin in maintaining the plasma membrane localizations and functions of human blood-brain barrier transporters.
Distinct roles of ezrin, radixin and moesin in maintaining the plasma membrane localizations and functions of human blood-brain barrier transporters.
Journal of cerebral blood flow and metabolism : official journal of the International Society of Cerebral Blood Flow and Metabolism (2019-08-15)
Yutaro Hoshi, Yasuo Uchida, Takashi Kuroda, Masanori Tachikawa, Pierre-Olivier Couraud, Takashi Suzuki, Tetsuya Terasaki
ABSTRACT
The purpose of this study was to clarify the roles of ERM proteins (ezrin/radixin/moesin) in the regulation of membrane localization and transport activity of transporters at the human blood-brain barrier (BBB). Ezrin or moesin knockdown in a human in vitro BBB model cell line (hCMEC/D3) reduced both BCRP and GLUT1 protein expression levels on the plasma membrane. Radixin knockdown reduced not only BCRP and GLUT1, but also P-gp membrane expression. These results indicate that P-gp, BCRP and GLUT1 proteins are maintained on the plasma membrane via different ERM proteins. Furthermore, moesin knockdown caused the largest decrease of P-gp and BCRP efflux activity among the ERM proteins, whereas GLUT1 influx activity was similarly reduced by knockdown of each ERM protein. To investigate how moesin knockdown reduced P-gp efflux activity without loss of P-gp from the plasma membrane, we examined the role of PKCβI. PKCβI increased P-gp phosphorylation and reduced P-gp efflux activity. Radixin and moesin proteins were detected in isolated human brain capillaries, and their protein abundances were within a 3-fold range, compared with those in hCMEC/D3 cell line. These findings may mean that ezrin, radixin and moesin maintain the functions of different transporters in different ways at the human BBB.
Sigma-Aldrich
MISSION® esiRNA, targeting human MSN
Sigma-Aldrich
MISSION® esiRNA, targeting human RDX
Sigma-Aldrich
Dantrolene sodium salt