Merck
  • The gut microbiota regulates autism-like behavior by mediating vitamin B6 homeostasis in EphB6-deficient mice.
The gut microbiota regulates autism-like behavior by mediating vitamin B6 homeostasis in EphB6-deficient mice.
Microbiome (2020-08-21)
Ying Li, Zheng-Yi Luo, Yu-Ying Hu, Yue-Wei Bi, Jian-Ming Yang, Wen-Jun Zou, Yun-Long Song, Shi Li, Tong Shen, Shu-Ji Li, Lang Huang, Ai-Jun Zhou, Tian-Ming Gao, Jian-Ming Li
ABSTRACT
Autism spectrum disorder (ASD) is a developmental disorder, and the effective pharmacological treatments for the core autistic symptoms are currently limited. Increasing evidence, particularly that from clinical studies on ASD patients, suggests a functional link between the gut microbiota and the development of ASD. However, the mechanisms linking the gut microbiota with brain dysfunctions (gut-brain axis) in ASD have not yet been full elucidated. Due to its genetic mutations and downregulated expression in patients with ASD, EPHB6, which also plays important roles in gut homeostasis, is generally considered a candidate gene for ASD. Nonetheless, the role and mechanism of EPHB6 in regulating the gut microbiota and the development of ASD are unclear. Here, we found that the deletion of EphB6 induced autism-like behavior and disturbed the gut microbiota in mice. More importantly, transplantation of the fecal microbiota from EphB6-deficient mice resulted in autism-like behavior in antibiotic-treated C57BL/6J mice, and transplantation of the fecal microbiota from wild-type mice ameliorated the autism-like behavior in EphB6-deficient mice. At the metabolic level, the disturbed gut microbiota in EphB6-deficient mice led to vitamin B6 and dopamine defects. At the cellular level, the excitation/inhibition (E/I) balance in the medial prefrontal cortex was regulated by gut microbiota-mediated vitamin B6 in EphB6-deficient mice. Our study uncovers a key role for the gut microbiota in the regulation of autism-like social behavior by vitamin B6, dopamine, and the E/I balance in EphB6-deficient mice, and these findings suggest new strategies for understanding and treating ASD. Video abstract.
Sigma-Aldrich
Fluorescein isothiocyanate–dextran, average mol wt 4,000, (FITC:Glucose = 1:250)
Serine, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Anti-c-Fos Antibody, clone 2G9C3, clone 2G9C3, from mouse
Sigma-Aldrich
Pyridoxal 5′-phosphate monohydrate, ≥97.0% (NT)
Sigma-Aldrich
Serotonin hydrochloride, powder
Sigma-Aldrich
Pyridoxine, ≥98%
Supelco
Serotonin, analytical standard