Merck
  • Th2 cytokines associated with chronic rhinosinusitis with polyps down-regulate the antimicrobial immune function of human sinonasal epithelial cells.
Th2 cytokines associated with chronic rhinosinusitis with polyps down-regulate the antimicrobial immune function of human sinonasal epithelial cells.
American journal of rhinology (2008-04-18)
Murugappan Ramanathan, Won-Kyung Lee, Ernst W Spannhake, Andrew P Lane
ABSTRACT
Chronic rhinosinusitis with nasal polyps (CRSwNPs) is a disorder characterized by persistent eosinophilic Th2 inflammation and frequent sinonasal microbial colonization. It has been postulated that an abnormal mucosal immune response underlies disease pathogenesis. The relationship between Th2 inflammatory cytokines and the innate immune function of sinonasal epithelial cells (SNECs) has not been explored. Human SNECs (HSNECs) isolated from control subjects and patients with CRS were assessed for expression of antimicrobial innate immune genes and proinflammatory cytokine genes by real-time polymerase chain reaction, ELISA, and flow cytometry. A model of the Th2 inflammatory environment was created by exposure of primary HSNEC to the Th2 cytokine interleukin (IL)-4 or IL-13 for 36 hours, with subsequent assessment of innate immune gene expression. HSNEC obtained from CRSwNP patients displayed decreased expression of multiple antimicrobial innate immune markers, including toll-like receptor 9, human beta-defensin 2, and surfactant protein A. Baseline expression of these genes by normal and CRS HSNEC in culture is significantly down-regulated after incubation with IL-4 or IL-13. Expression of multiple innate immune genes by HSNEC is reduced in CRSwNP. One mechanism appears to be a direct effect of the leukocyte-derived Th2 cytokines present in the sinonasal mucosa in CRSwNP. Impaired mucosal innate immunity may contribute to microbial colonization and abnormal immune responses associated with CRSwNP.
Sigma-Aldrich
Polyvinylpyrrolidone, K 30
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Polyvinylpyrrolidone, average Mw ~1,300,000 by LS
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Polyvinylpyrrolidone, K 90
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Polyvinylpyrrolidone, powder, average Mw ~55,000
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Polyvinylpyrrolidone, powder, average Mw ~29,000
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Polyvinylpyrrolidone, suitable for plant cell culture, average mol wt 10,000
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Polyvinylpyrrolidone, powder, BioXtra, suitable for mouse embryo cell culture
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Polyvinylpyrrolidone, mol wt (number average molecular weight Mn 360kDa)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Polyvinylpyrrolidone, average mol wt 10,000
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Polyvinylpyrrolidone, average mol wt 40,000
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Polyvinylpyrrolidone, for molecular biology, nucleic acid hybridization tested, mol wt 360,000
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Collagen Type IV from human cell culture, Bornstein and Traub Type IV, 0.3 mg/mL, sterile-filtered, BioReagent, suitable for cell culture