Merck
  • Artificially decreasing cortical tension generates aneuploidy in mouse oocytes.
Artificially decreasing cortical tension generates aneuploidy in mouse oocytes.
Nature communications (2020-04-05)
Isma Bennabi, Flora Crozet, Elvira Nikalayevich, Agathe Chaigne, Gaëlle Letort, Marion Manil-Ségalen, Clément Campillo, Clotilde Cadart, Alice Othmani, Rafaele Attia, Auguste Genovesio, Marie-Hélène Verlhac, Marie-Emilie Terret
ABSTRACT
Human and mouse oocytes' developmental potential can be predicted by their mechanical properties. Their development into blastocysts requires a specific stiffness window. In this study, we combine live-cell and computational imaging, laser ablation, and biophysical measurements to investigate how deregulation of cortex tension in the oocyte contributes to early developmental failure. We focus on extra-soft cells, the most common defect in a natural population. Using two independent tools to artificially decrease cortical tension, we show that chromosome alignment is impaired in extra-soft mouse oocytes, despite normal spindle morphogenesis and dynamics, inducing aneuploidy. The main cause is a cytoplasmic increase in myosin-II activity that could sterically hinder chromosome capture. We describe here an original mode of generation of aneuploidies that could be very common in oocytes and could contribute to the high aneuploidy rate observed during female meiosis, a leading cause of infertility and congenital disorders.
Sigma-Aldrich
Nile Red, Technical grade
Sigma-Aldrich
ML-7, powder
Sigma-Aldrich
Monastrol, ≥98% (HPLC), solid
Sigma-Aldrich
ML-7, Hydrochloride, A cell-permeable, potent, reversible, ATP-competitive, and selective inhibitor of myosin light chain kinase (Ki = 300 nM).
Sigma-Aldrich
Fluorescein isothiocyanate–dextran, average mol wt 60,000-76,000
Sigma-Aldrich
U0126, U0126, CAS 109511-58-2, is a potent and specific inhibitor of MEK1 (IC₅₀ = 72 nM) and MEK2 (IC₅₀ = 58 nM). The inhibition is noncompetitive with respect to both ATP and ERK.