Merck
  • Interleukin-4 and interleukin-13 inhibit the expression of leukemia inhibitory factor and interleukin-11 in fibroblasts.
Interleukin-4 and interleukin-13 inhibit the expression of leukemia inhibitory factor and interleukin-11 in fibroblasts.
Molecular immunology (2011-12-07)
Pedro P C Souza, Py Palmqvist, Pernilla Lundberg, Inger Lundgren, Lennart Hänström, João A C Souza, H Herschel Conaway, Ulf H Lerner
ABSTRACT
Cytokines produced by inflammatory or resident mesenchymal cells play important modulatory roles in the pathogenesis of inflammation induced bone loss. In the present study, the effects of IL-4 and IL-13 on the expression of three osteotropic cytokines in the IL-6 family expressed in human gingival fibroblasts were studied. IL-4Rα and IL-13Rα1 mRNA were constitutively expressed in human gingival fibroblasts. The inflammatory cytokines IL-1β and TNF-α increased expression of IL-6, LIF, and IL-11 mRNA and protein in the gingival fibroblasts. Addition of IL-4 or IL-13 had no effect on IL-6 expression, but significantly inhibited LIF and IL-11 mRNA and protein stimulated by IL-1β and TNF-α. No involvement of NF-κB or STAT1 was observed in the inhibition. STAT6 was phosphorylated at Y641 by treatment with IL-4 and knockdown of STAT6 with siRNA decreased the inhibition of IL-11 and LIF expression by IL-4 in IL-1β and TNF-α stimulated cells. This study suggests that activation of STAT6 by IL-4 and IL-13, through type 2 IL-4 receptors, inhibits production of IL-11 and LIF stimulated by IL-1β and TNF-α in human gingival fibroblasts. A negative modulatory role of IL-4 and IL-13 in osteotropic cytokine production could be a mechanism playing an important inhibitory role in inflammation induced periodontitis.
Product Description
Roche
KAPA2G Robust HotStart ReadyMix, 2 ×, with dye