Merck
  • G/T substitution in intron 1 of the UNC13B gene is associated with increased risk of nephropathy in patients with type 1 diabetes.
G/T substitution in intron 1 of the UNC13B gene is associated with increased risk of nephropathy in patients with type 1 diabetes.
Diabetes (2008-07-18)
David-Alexandre Trégouet, Per-Henrik Groop, Steven McGinn, Carol Forsblom, Samy Hadjadj, Michel Marre, Hans-Henrik Parving, Lise Tarnow, Ralph Telgmann, Tiphaine Godefroy, Viviane Nicaud, Rachel Rousseau, Maikki Parkkonen, Anna Hoverfält, Ivo Gut, Simon Heath, Fumihiko Matsuda, Roger Cox, Gbenga Kazeem, Martin Farrall, Dominique Gauguier, Stefan-Martin Brand-Herrmann, François Cambien, Mark Lathrop, Nathalie Vionnet
ABSTRACT
Genetic and environmental factors modulate the susceptibility to diabetic nephropathy, as initiating and/or progression factors. The objective of the European Rational Approach for the Genetics of Diabetic Complications (EURAGEDIC) study is to identify nephropathy susceptibility genes. We report molecular genetic studies for 127 candidate genes for nephropathy. Polymorphisms were identified through sequencing of promoter, exon, and flanking intron gene regions and a database search. A total of 344 nonredundant SNPs and nonsynonymous variants were tested for association with diabetic nephropathy (persistent albuminuria >/=300 mg/24 h) in a large type 1 diabetes case/control (1,176/1,323) study from three European populations. Only one SNP, rs2281999, located in the UNC13B gene, was significantly associated with nephropathy after correction for multiple testing. Analyses of 21 additional markers fully characterizing the haplotypic variability of the UNC13B gene showed consistent association of SNP rs13293564 (G/T) located in intron 1 of the gene with nephropathy in the three populations. The odds ratio (OR) for nephropathy associated with the TT genotype was 1.68 (95% CI 1.29-2.19) (P = 1.0 x 10(-4)). This association was replicated in an independent population of 412 case subjects and 614 control subjects (combined OR of 1.63 [95% CI 1.30-2.05], P = 2.3 x 10(-5)). We identified a polymorphism in the UNC13B gene associated with nephropathy. UNC13B mediates apopotosis in glomerular cells in the presence of hyperglycemia, an event occurring early in the development of nephropathy. We propose that this polymorphism could be a marker for the initiation of nephropathy. However, further studies are needed to clarify the role of UNC13B in nephropathy.
Sigma-Aldrich
StableCell RPMI-1640, With stable glutamine and sodium bicarbonate, liquid, sterile-filtered, suitable for cell culture
SAFC
RPMI-1640 Medium, with 2.05 mM L-glutamine, with 25mM HEPES, liquid, sterile-filtered, suitable for cell culture
SAFC
RPMI-1640 Medium, HEPES Modification, With 25 mM HEPES, without L-glutamine., liquid, sterile-filtered, suitable for cell culture
Sigma-Aldrich
RPMI-1640 Medium, Modified, with sodium bicarbonate, without L-glutamine and phenol red, liquid, sterile-filtered, suitable for cell culture
Sigma-Aldrich
RPMI-1640 Medium, With L-glutamine, without sodium bicarbonate, powder, suitable for cell culture
Sigma-Aldrich
RPMI-1640 Medium, Modified, with 20 mM HEPES and L-glutamine, without sodium bicarbonate, liquid, sterile-filtered, suitable for cell culture
Sigma-Aldrich
RPMI-1640 Medium, Modified, with L-glutamine, without phenol red and sodium bicarbonate, powder, suitable for cell culture
SAFC
RPMI-1640 Medium, HEPES Modification, with L-glutamine and 25mM HEPES, without sodium bicarbonate, powder, suitable for cell culture
Sigma-Aldrich
RPMI-1640 Medium, With L-glutamine and sodium bicarbonate, liquid, sterile-filtered, suitable for cell culture
Sigma-Aldrich
RPMI-1640 Medium, Dutch Modification, with sodium bicarbonate and 20mM HEPES, without L-glutamine, liquid, sterile-filtered, suitable for cell culture
SAFC
RPMI-1640 Medium, AutoMod, without L-glutamine and sodium bicarbonate, powder, suitable for cell culture
Sigma-Aldrich
RPMI-1640 Medium, With L-glutamine, without glucose and sodium bicarbonate, powder, suitable for cell culture
Sigma-Aldrich
RPMI-1640 Medium, With L-glutamine and sodium bicarbonate. Without arginine, leucine, lysine, and phenol red, liquid, sterile-filtered, suitable for cell culture, designed for isotope labeling for cell culture applications
SAFC
RPMI-1640 Medium, 10 ×, Without L-glutamine, folic acid and sodium bicarbonate, liquid, sterile-filtered, suitable for cell culture
Sigma-Aldrich
RPMI-1640 Medium, Modified, with sodium bicarbonate, without methionine, cystine and L-glutamine, liquid, sterile-filtered, suitable for cell culture
SAFC
RPMI-1640 Medium, Hybri-Max, Modified, with L-glutamine, 4500 mg/L glucose and 15mM HEPES, without sodium bicarbonate, powder, suitable for hybridoma
Sigma-Aldrich
RPMI-1640 Medium, With sodium bicarbonate, without L-glutamine, liquid, sterile-filtered, suitable for cell culture