Merck
  • Induction of apoptosis in human cancer cells by candidaspongiolide, a novel sponge polyketide.
Induction of apoptosis in human cancer cells by candidaspongiolide, a novel sponge polyketide.
Journal of the National Cancer Institute (2008-08-30)
Daniela Trisciuoglio, Badarch Uranchimeg, John H Cardellina, Tamara L Meragelman, Shigeki Matsunaga, Nobuhiru Fusetani, Donatella Del Bufalo, Robert H Shoemaker, Giovanni Melillo
ABSTRACT
Candidaspongiolide (CAN), a novel polyketide from a marine sponge, is the active component of a mixture that was found to be potently cytotoxic in the National Cancer Institute's 60-cell-line screen. Effects of CAN on U251 glioma and HCT116 colorectal cancer cells and on normal fibroblasts were assessed using radiolabeling studies to measure protein synthesis, clonogenic assays to measure cell survival, flow cytometry of annexin V- and propidium iodide-stained cells to measure apoptosis, and western blots in the presence or absence of specific inhibitors to assess accumulation and phosphorylation of potential downstream target proteins. CAN inhibited protein synthesis and potently induced apoptosis in both U251 and HCT116 cells, the latter in part by a caspase 12-dependent pathway. For example, 25%-30% of U251 or HCT116 cells became apoptotic after 24 hours of treatment with 100 nM CAN. CAN also rapidly induced sustained phosphorylation of eukaryotic translation initiation factor-2 (eIF2)-alpha at Ser51 and of the translation elongation factor eEF2 at Thr56, which could contribute to its dose-dependent inhibition of protein synthesis. Stable expression of dominant-negative eIF2alpha was sufficient to prevent CAN-induced eIF2alpha phosphorylation and induction of apoptosis but insufficient to prevent inhibition of protein synthesis. CAN induction of eIF2alpha phosphorylation did not occur by a classic endoplasmic reticulum stress pathway. However, an inhibitor of and small-interfering RNAs to the double-stranded RNA-dependent protein kinase PKR prevented CAN-mediated eIF2alpha phosphorylation and apoptosis, respectively. Although CAN inhibited protein synthesis in both cancer cells and normal human fibroblasts, it induced eIF2alpha phosphorylation and apoptosis only in cancer cells. CAN triggers PKR/eIF2alpha/caspase 12-dependent apoptosis and inhibits protein synthesis in cancer cells but only inhibits protein synthesis in normal cells.
Sigma-Aldrich
Crystal Violet, S. No.: 785
Supelco
Crystal Violet, VETRANAL®, analytical standard
Sigma-Aldrich
Crystal Violet, certified by the BSC
Sigma-Aldrich
Crystal Violet, ACS reagent, ≥90.0% anhydrous basis
Sigma-Aldrich
Crystal Violet
Sigma-Aldrich
(S)-(+)-Camptothecin, ≥90% (HPLC), powder