Merck
  • Regenerative Metaplastic Clones in COPD Lung Drive Inflammation and Fibrosis.
Regenerative Metaplastic Clones in COPD Lung Drive Inflammation and Fibrosis.
Cell (2020-04-17)
Wei Rao, Shan Wang, Marcin Duleba, Suchan Niroula, Kristina Goller, Jingzhong Xie, Rajasekaran Mahalingam, Rahul Neupane, Audrey-Ann Liew, Matthew Vincent, Kenichi Okuda, Wanda K O'Neal, Richard C Boucher, Burton F Dickey, Michael E Wechsler, Omar Ibrahim, John F Engelhardt, Tinne C J Mertens, Wei Wang, Soma S K Jyothula, Christopher P Crum, Harry Karmouty-Quintana, Kalpaj R Parekh, Mark L Metersky, Frank D McKeon, Wa Xian
ABSTRACT
Chronic obstructive pulmonary disease (COPD) is a progressive condition of chronic bronchitis, small airway obstruction, and emphysema that represents a leading cause of death worldwide. While inflammation, fibrosis, mucus hypersecretion, and metaplastic epithelial lesions are hallmarks of this disease, their origins and dependent relationships remain unclear. Here we apply single-cell cloning technologies to lung tissue of patients with and without COPD. Unlike control lungs, which were dominated by normal distal airway progenitor cells, COPD lungs were inundated by three variant progenitors epigenetically committed to distinct metaplastic lesions. When transplanted to immunodeficient mice, these variant clones induced pathology akin to the mucous and squamous metaplasia, neutrophilic inflammation, and fibrosis seen in COPD. Remarkably, similar variants pre-exist as minor constituents of control and fetal lung and conceivably act in normal processes of immune surveillance. However, these same variants likely catalyze the pathologic and progressive features of COPD when expanded to high numbers.
Sigma-Aldrich
Triton X-100, laboratory grade
Sigma-Aldrich
Anti-IVL antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Anti-SFTPB antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Anti-Acetylated Tubulin antibody, Mouse monoclonal, clone 6-11B-1, purified from hybridoma cell culture
Sigma-Aldrich
Anti-AQP4 antibody produced in rabbit, affinity isolated antibody, Prestige Antibodies® Powered by Atlas Antibodies, buffered aqueous glycerol solution