Merck
  • Liver and kidney serum profile abnormalities in workers exposed to aflatoxin B1 in urban solid waste management centers.
Liver and kidney serum profile abnormalities in workers exposed to aflatoxin B1 in urban solid waste management centers.
Environmental monitoring and assessment (2020-07-02)
Morvarid Karamkhani, Hassan Asilian-Mahabadi, Bahram Daraei, Ali Seidkhani-Nahal, Ali Noori-Zadeh
ABSTRACT
Many workers are exposed to health problems arising from molds, fungi, and their toxins during waste processing. Aflatoxin B1 (AFB1) level in airborne and settled dust, aflatoxin B1-albumin (AFB1-Alb) adduct in serum, liver and kidney biochemical tests, and body redox change of workers in municipal dry waste-processing sites were investigated. The surface, personal, and area air dust and the blood of workers' samples were collected from the plastic and bread waste-sorting sections in three recycling municipal dry waste sites. Digestion (only for serum samples), passed through SPE cartridge, elution, and collection with methanol, immune-affinity column clean-up, and HPLC system equipped with post-column derivatization method and fluorescence detection were performed for determination of AFB1 and AFB1-Alb levels in the samples. The mean level of dust and AFB1 in the personal and area air, and in the settled dust and the AFB1-Alb in the serum of workers in the bread waste sorting, was higher than plastic waste-sorting samples, in all of the sites. The differences in the biochemical profiles of subjects exposed to aflatoxin B1 as compared to the control group especially in liver and kidney function parameters as well as antioxidant factors of the serum were significant. The workers in handling of municipal waste may be exposed to potentially hazardous levels of aflatoxin B1. The adverse effects of AFB1 on the kidney and liver may be caused by changes in the redox system.
Sigma-Aldrich
Aflatoxin B1−BSA Conjugate from Aspergillus flavus
Pricing and availability is not currently available.