Merck
  • Development of novel and green NiFe2O4/geopolymer nanocatalyst based on bentonite for synthesis of imidazole heterocycles by ultrasonic irradiations.
Development of novel and green NiFe2O4/geopolymer nanocatalyst based on bentonite for synthesis of imidazole heterocycles by ultrasonic irradiations.
Scientific reports (2020-07-17)
Zoleikha Hajizadeh, Fateme Radinekiyan, Reza Eivazzadeh-Keihan, Ali Maleki
ABSTRACT
Geopolymers as aluminosilicate inorganic polymers and eco-friendly building materials which can be used as substrate for different kinds of composite. In this research, according to the fabrication of geopolymer based on bentonite as a substrate and embedment of NiFe2O4 nanoparticles in the construction of this polymer, the synthesis of a new magnetic nanocomposite (NiFe2O4/geopolymer) was investigated for the first time. In order to describe its chemistry and morphology features, different analyses such as Fourier transform infrared spectroscopy, field-emission scanning electron microscopy and transmission electron microscopy images, Brunauer-Emmet-Teller adsorption-desorption isotherm, X-ray diffraction pattern, energy-dispersive X-ray analysis, thermogravimetric analysis, and vibrating-sample magnetometer analysis were used. The application of this novel nanocatalyst was studied for one-pot three-component condensation reaction of substituted imidazole derivatives by accelerated ultrasonic irradiations. Compared to the other conventional catalysts which were used for the synthesis of imidazole derivatives, the green synthesis method for fabrication of this heterogeneous and magnetic nanocatalyst, its high thermal stability, being eco-friendly, noticeable efficiency and easy reusability have become privileges to be superior.