Merck
  • Illumination guidelines for ultrafast pump-probe experiments by serial femtosecond crystallography.
Illumination guidelines for ultrafast pump-probe experiments by serial femtosecond crystallography.
Nature methods (2020-05-27)
Marie Luise Grünbein, Miriam Stricker, Gabriela Nass Kovacs, Marco Kloos, R Bruce Doak, Robert L Shoeman, Jochen Reinstein, Sylvain Lecler, Stefan Haacke, Ilme Schlichting
ABSTRACT
Time-resolved crystallography with X-ray free-electron lasers enables structural characterization of light-induced reactions on ultrafast timescales. To be biologically and chemically relevant, such studies must be carried out in an appropriate photoexcitation regime to avoid multiphoton artifacts, a common issue in recent studies. We describe numerical and experimental approaches to determine how many photons are needed for single-photon excitation in microcrystals, taking into account losses by scattering.
Sigma-Aldrich
Agarose, low gelling temperature, BioReagent, for molecular biology
Sigma-Aldrich
Pluronic® F-127, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Carboxymethylcellulose sodium salt, ultra high viscosity, highly purified