Merck
  • Inducible Germline IgMs Bridge Trypanosome Lytic Factor Assembly and Parasite Recognition.
Inducible Germline IgMs Bridge Trypanosome Lytic Factor Assembly and Parasite Recognition.
Cell host & microbe (2020-05-18)
Joseph Verdi, Ronnie Zipkin, Elani Hillman, Rahel A Gertsch, Sarah J Pangburn, Russell Thomson, Nina Papavasiliou, Jeremy Sternberg, Jayne Raper
ABSTRACT
Trypanosomiasis is a devastating neglected tropical disease affecting livestock and humans. Humans are susceptible to two Trypanosoma brucei subspecies but protected from other trypanosomes by circulating high-density lipoprotein (HDL) complexes called trypanosome lytic factors (TLFs) 1 and 2. TLFs contain apolipoprotein L-1 contributing to lysis and haptoglobin-related protein (HPR), which can function as a ligand for a parasite receptor. TLF2 also uniquely contains non-covalently associated immunoglobin M (IgM) antibodies, the role and origin of which remain unclear. Here, we show that these TLF2-associated IgMs interact with both HPR and alternate trypanosome surface proteins, including variant surface glycoprotein, likely facilitating complex biogenesis and TLF uptake into parasites. TLF2-IgMs are germline antibodies that, while present at basal concentrations in healthy individuals, are elicited by trypanosome infection in both murine models and human sleeping sickness patients. These data suggest that poly- and self-reactive germline antibodies such as TLF2-associated IgMs play a role in antimicrobial immunity.
Product Description
Supelco
Diminazene aceturate, analytical standard
Sigma-Aldrich
L-Cysteine, from non-animal source, BioReagent, suitable for cell culture, ≥98%
Sigma-Aldrich
IgM from human serum, reagent grade, ~95% (HPLC), buffered aqueous solution
Sigma-Aldrich
Hypoxanthine, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Monoclonal Anti-Haptoglobin antibody produced in mouse, clone HG-36, ascites fluid