Merck
  • Endonuclease FEN1 Coregulates ERα Activity and Provides a Novel Drug Interface in Tamoxifen-Resistant Breast Cancer.
Endonuclease FEN1 Coregulates ERα Activity and Provides a Novel Drug Interface in Tamoxifen-Resistant Breast Cancer.
Cancer research (2020-03-21)
Koen D Flach, Manikandan Periyasamy, Ajit Jadhav, Dorjbal Dorjsuren, Joseph C Siefert, Theresa E Hickey, Mark Opdam, Hetal Patel, Sander Canisius, David M Wilson, Maria Donaldson Collier, Stefan Prekovic, Marja Nieuwland, Roelof J C Kluin, Alexey V Zakharov, Jelle Wesseling, Lodewyk F A Wessels, Sabine C Linn, Wayne D Tilley, Anton Simeonov, Simak Ali, Wilbert Zwart
ABSTRACT
Estrogen receptor α (ERα) is a key transcriptional regulator in the majority of breast cancers. ERα-positive patients are frequently treated with tamoxifen, but resistance is common. In this study, we refined a previously identified 111-gene outcome prediction-classifier, revealing FEN1 as the strongest determining factor in ERα-positive patient prognostication. FEN1 levels were predictive of outcome in tamoxifen-treated patients, and FEN1 played a causal role in ERα-driven cell growth. FEN1 impacted the transcriptional activity of ERα by facilitating coactivator recruitment to the ERα transcriptional complex. FEN1 blockade induced proteasome-mediated degradation of activated ERα, resulting in loss of ERα-driven gene expression and eradicated tumor cell proliferation. Finally, a high-throughput 465,195 compound screen identified a novel FEN1 inhibitor, which effectively blocked ERα function and inhibited proliferation of tamoxifen-resistant cell lines as well as ex vivo-cultured ERα-positive breast tumors. Collectively, these results provide therapeutic proof of principle for FEN1 blockade in tamoxifen-resistant breast cancer. SIGNIFICANCE: These findings show that pharmacologic inhibition of FEN1, which is predictive of outcome in tamoxifen-treated patients, effectively blocks ERα function and inhibits proliferation of tamoxifen-resistant tumor cells.
Sigma-Aldrich
Retinoic acid, ≥98% (HPLC), powder
Sigma-Aldrich
Anti-phospho-Histone H2A.X (Ser139) Antibody, clone JBW301, clone JBW301, Upstate®, from mouse
Sigma-Aldrich
Anti-Actin Antibody, clone C4, clone C4, Chemicon®, from mouse
Sigma-Aldrich
3,3′-Diaminobenzidine tetrahydrochloride hydrate, ISOPAC®
Sigma-Aldrich
MISSION® esiRNA, targeting human FEN1