Merck
  • Molecular Transducers of Human Skeletal Muscle Remodeling under Different Loading States.
Molecular Transducers of Human Skeletal Muscle Remodeling under Different Loading States.
Cell reports (2020-08-07)
Tanner Stokes, James A Timmons, Hannah Crossland, Thomas R Tripp, Kevin Murphy, Chris McGlory, Cameron J Mitchell, Sara Y Oikawa, Robert W Morton, Bethan E Phillips, Steven K Baker, Phillip J Atherton, Claes Wahlestedt, Stuart M Phillips
ABSTRACT
Loading of skeletal muscle changes the tissue phenotype reflecting altered metabolic and functional demands. In humans, heterogeneous adaptation to loading complicates the identification of the underpinning molecular regulators. A within-person differential loading and analysis strategy reduces heterogeneity for changes in muscle mass by ∼40% and uses a genome-wide transcriptome method that models each mRNA from coding exons and 3' and 5' untranslated regions (UTRs). Our strategy detects ∼3-4 times more regulated genes than similarly sized studies, including substantial UTR-selective regulation undetected by other methods. We discover a core of 141 genes correlated to muscle growth, which we validate from newly analyzed independent samples (n = 100). Further validating these identified genes via RNAi in primary muscle cells, we demonstrate that members of the core genes were regulators of protein synthesis. Using proteome-constrained networks and pathway analysis reveals notable relationships with the molecular characteristics of human muscle aging and insulin sensitivity, as well as potential drug therapies.
Sigma-Aldrich
Puromycin dihydrochloride from Streptomyces alboniger, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
MISSION® esiRNA, targeting human FKBP1A
Sigma-Aldrich
MISSION® esiRNA, targeting human NID2
Sigma-Aldrich
MISSION® esiRNA, targeting human BCAT2
Sigma-Aldrich
MISSION® esiRNA, targeting human MBNL1
Sigma-Aldrich
Anti-Puromycin Antibody, clone 12D10, clone 12D10, from mouse
Roche
cOmplete, Mini, EDTA-free Protease Inhibitor Cocktail, Protease Inhibitor Cocktail Tablets provided in a glass vial, Tablets provided in a glass vial