Merck
  • Hedgehog signaling pathway regulates gene expression profile of epididymal principal cells through the primary cilium.
Hedgehog signaling pathway regulates gene expression profile of epididymal principal cells through the primary cilium.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology (2020-04-14)
Laura Girardet, Agathe Bernet, Ezéquiel Calvo, Denis Soulet, Charles Joly-Beauparlant, Arnaud Droit, Daniel G Cyr, Clémence Belleannée
ABSTRACT
Primary cilia (PC) are organelles that sense and respond to dynamic changes of the extracellular milieu through the regulation of target genes. By using the epididymis as a model system, we determined the contribution of primary cilia in the regulation of epithelial cell functions through the transduction of the Hedgehog (Hh) signaling pathway. Both Sonic (SHH) and Indian Hedgehog (IHH) ligands were detected in epididymal epithelial cells by confocal microscopy and found secreted in the extracellular space. Gene expression profiling preformed on ciliated epithelial cells indicated that 153 and 1052 genes were differentially expressed following treatment with the Hh agonist SAG or the Hh antagonist cyclopamine (Cyclo), respectively. Strikingly, gene ontology analysis indicated that genes associated with immune response were the most affected following Hh modulation. The contribution of epididymal PC to canonical Hh pathway transduction was validated by ciliobrevin D treatment, which induced a significant decrease in PC length and a reduction in the expression Hh signaling targets. Such findings bring us closer to a molecular understanding of the subtle immune balance observed in some epithelia, including the epididymis and the intestine, which are organs featuring both tolerance toward autoimmune spermatozoa (or commensal bacteria) and defense against pathogens.
Sigma-Aldrich
Smoothened Agonist, SAG, A cell-permeable Smoothened Agonist, SAG, CAS 364590-63-6, modulates the coupling of Smo with its downstream effector by interacting with the Smo heptahelical domain (KD = 59 nM).
Sigma-Aldrich
Human Plasma Fibronectin Purified Protein, from human plasma, liquid, 1 mg/mL (100 MG pack size is lyophilized), purified protein, suitable for cell culture
Sigma-Aldrich
2-Mercaptoethanol, for molecular biology, suitable for electrophoresis, suitable for cell culture, BioReagent, 99% (GC/titration)
Sigma-Aldrich
Lipopolysaccharides from Escherichia coli O111:B4, γ-irradiated, BioXtra, suitable for cell culture