Merck
  • A supramolecular platform for controlling and optimizing molecular architectures of siRNA targeted delivery vehicles.
A supramolecular platform for controlling and optimizing molecular architectures of siRNA targeted delivery vehicles.
Science advances (2020-08-25)
Yuting Wen, Hongzhen Bai, Jingling Zhu, Xia Song, Guping Tang, Jun Li
ABSTRACT
It requires multistep synthesis and conjugation processes to incorporate multifunctionalities into a polyplex gene vehicle to overcome numerous hurdles during gene delivery. Here, we describe a supramolecular platform to precisely control, screen, and optimize molecular architectures of siRNA targeted delivery vehicles, which is based on rationally designed host-guest complexation between a β-cyclodextrin-based cationic host polymer and a library of guest polymers with various PEG shape and size, and various density of ligands. The host polymer is responsible to load/unload siRNA, while the guest polymer is responsible to shield the vehicles from nonspecific cellular uptake, to prolong their circulation time, and to target tumor cells. A series of precisely controlled molecular architectures through a simple assembly process allow for a rapid optimization of siRNA delivery vehicles in vitro and in vivo for therapeutic siRNA-Bcl2 delivery and tumor therapy, indicating the platform is a powerful screening tool for targeted gene delivery vehicles.
Sigma-Aldrich
MISSION® esiRNA, targeting human BCL2
Pricing and availability is not currently available.
Sigma-Aldrich
2-Phenylindole, technical grade, 95%
Pricing and availability is not currently available.
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Bcl2
Pricing and availability is not currently available.