Merck
  • Preparation of N2-Ethyl-2'-deoxyguanosine-d4 as an Internal Standard for the Electrospray Ionization-Tandem Mass Spectrometric Determination of DNA Damage by Acetaldehyde.
Preparation of N2-Ethyl-2'-deoxyguanosine-d4 as an Internal Standard for the Electrospray Ionization-Tandem Mass Spectrometric Determination of DNA Damage by Acetaldehyde.
Analytical sciences : the international journal of the Japan Society for Analytical Chemistry (2020-01-28)
Yukihiro Esaka, Hiromitsu Aruga, Saki Kunishima, Takuhei Yamamoto, Hiroya Murakami, Yoshinari Sawama, Hironao Sajiki, Bunji Uno
ABSTRACT
The deuteration of N2-ethyl-2'-deoxyguanosine (Et-dG), which is a DNA adduct generated from acetaldehyde, was studied by the addition reaction of acetaldehyde-d4 to 2'-deoxyguanosine (dG) in deuterium oxide (D2O), with the aim to obtain an isotope internal standard for the liquid chromatography/tandem mass spectrometry (LC/MS/MS) quantitation of Et-dG. The replacement of the dG C-8 hydrogen atom by a deuteron atom took place at 50°C in D2O and afforded a mixture of Et-dG-d4 and Et-dG-d5. Et-dG-d4, which was stable in aqueous solutions, was prepared by incubating the mixture in H2O at 60°C for 48 h. The calibration curve was obtained by multiple reaction monitoring (MRM) measurements using a hydrophilic interaction chromatography-electrospray ionization-tandem mass spectrometric (HILIC/ESI-MS/MS) system between the Et-dG concentration, ranging from 1.0 × 10-10 to 4.0 × 10-9 M in the sample solutions, and the relative peak areas of Et-dG (m/z: 296.1 → 180.1) to the value of Et-dG-d4 (m/z: 300.2 → 184.2), with an internal standard showing good linearity (R2 = 0.995, n = 5).
Sigma-Aldrich
Acetaldehyde-d4, ≥99 atom % D, ≥98% (CP)