Merck
X6: A Novel Antibody for Potential Use in Gluten Quantification.
Molecules (Basel, Switzerland) (2020-07-12)
Aleksandrina Shatalova, Ivan Shatalov, Yuri Lebedin
ABSTRACT
Gliadin is a fraction of gluten, known to trigger celiac disease in susceptible people. To date, the life-long gluten-free diet is used for the prevention of this disease. Hence, methods for gluten control in foods are of significant importance. Being one of the most-used methods used for this purpose, ELISA should use high-affinity antibodies to gliadin peptides involved into celiac process. This study investigates the characteristics of a novel anti-gliadin antibody X6. We found the QXQPFPXP site to be a recognized epitope that provides specific binding of the antibody to cereal prolamins involved in celiac disease manifestation. A specificity study using immunoblotting shows the recognition of wheat, barley and rye proteins-as well as α-gliadin homologs from non-edible cereals (Dasypyrum villosum). Reactivity to avenin was less pronounced, as this protein does not contain the PFP motif most critical for antibody recognition. The proteins of Zea mays and Setaria italica were not recognized by X6. X6-based ELISA highly correlated with R5 and G12, which are Codex Alimentarius standards in the quantitative assessment of gluten content (Pearson's R = 0.86 and 0.87, respectively). Qualitative assessment revealed no significant differences between R5 and G12 and X6.
Sigma-Aldrich
Gliadin from wheat
Sigma-Aldrich
E-64d, protease inhibitor