Merck
  • Development and validation of a UHPLC-MS/MS method for quantification of the prodrug remdesivir and its metabolite GS-441524: a tool for clinical pharmacokinetics of SARS-CoV-2/COVID-19 and Ebola virus disease.
Development and validation of a UHPLC-MS/MS method for quantification of the prodrug remdesivir and its metabolite GS-441524: a tool for clinical pharmacokinetics of SARS-CoV-2/COVID-19 and Ebola virus disease.
The Journal of antimicrobial chemotherapy (2020-05-04)
Valeria Avataneo, Amedeo de Nicolò, Jessica Cusato, Miriam Antonucci, Alessandra Manca, Alice Palermiti, Catriona Waitt, Stephen Walimbwa, Mohammed Lamorde, Giovanni di Perri, Antonio D'Avolio
ABSTRACT
Remdesivir has received significant attention for its potential application in the treatment of COVID-19, caused by SARS-CoV-2. Remdesivir has already been tested for Ebola virus disease treatment and found to have activity against SARS and MERS coronaviruses. The remdesivir core contains GS-441524, which interferes with RNA-dependent RNA polymerases alone. In non-human primates, following IV administration, remdesivir is rapidly distributed into PBMCs and converted within 2 h to the active nucleoside triphosphate form, while GS-441524 is detectable in plasma for up to 24 h. Nevertheless, remdesivir pharmacokinetics and pharmacodynamics in humans are still unexplored, highlighting the need for a precise analytical method for remdesivir and GS-441524 quantification. The validation of a reliable UHPLC-MS/MS method for remdesivir and GS-441524 quantification in human plasma. Remdesivir and GS-441524 standards and quality controls were prepared in plasma from healthy donors. Sample preparation consisted of protein precipitation, followed by dilution and injection into the QSight 220 UHPLC-MS/MS system. Chromatographic separation was obtained through an Acquity HSS T3 1.8 μm, 2.1 × 50 mm column, with a gradient of water and acetonitrile with 0.05% formic acid. The method was validated using EMA and FDA guidelines. Analyte stability has been evaluated and described in detail. The method successfully fulfilled the validation process and it was demonstrated that, when possible, sample thermal inactivation could be a good choice in order to improve biosafety. This method represents a useful tool for studying remdesivir and GS-441524 clinical pharmacokinetics, particularly during the current COVID-19 outbreak.
Sigma-Aldrich
Atazanavir, ≥98% (HPLC)
Sigma-Aldrich
Amprenavir, ≥98% (HPLC)
Sigma-Aldrich
6,7-Dimethyl-2,3-di(2-pyridyl)quinoxaline, 98%
Sigma-Aldrich
Darunavir, ≥98% (HPLC)