Merck
A novel family of bromodomain genes.
Genomics (2000-02-09)
M H Jones, N Hamana, J i Nezu, M Shimane
ABSTRACT
The bromodomain is a structural motif characteristic of proteins involved in chromatin-dependent regulation of transcription. Bromodomain proteins have been identified as integral components of chromatin remodeling complexes and frequently possess histone acetyltransferase activity. Their encoding genes have been identified at translocation breakpoints, and at least one, CBP, is a tumor suppressor gene. We have identified a series of novel bromodomain genes by EST database and cDNA library screening. Comparison of sequences for four clones indicated that they represent genes belonging to a novel bromodomain family. Full-length sequences for these genes, which are widely expressed, predict encoded proteins of between 1527 and 1972 amino acids. In addition to a carboxy-terminal bromodomain, an adjacent PHD finger, and a WACZ motif, at least four other conserved novel motifs are present in each protein. The genes contain regions conserved with Drosophila Acf1 and Caenorhabditis elegans ZK783.4. The novel genes, termed BAZ1A, BAZ1B, BAZ2A, and BAZ2B, localize to chromosomes 14q12-q13, 7q11-q21, 12q24.3-qter, and 2q23-q24, respectively. Conservation of multiple domains throughout these genes with Acf1 indicates that they are likely to be components of chromatin remodeling complexes.
Sigma-Aldrich
BAZ2B (2054-2168) human, recombinant, expressed in E. coli, ≥84% (SDS-PAGE)