Merck
  • Preservation of microvascular barrier function requires CD31 receptor-induced metabolic reprogramming.
Preservation of microvascular barrier function requires CD31 receptor-induced metabolic reprogramming.
Nature communications (2020-07-19)
Kenneth C P Cheung, Silvia Fanti, Claudio Mauro, Guosu Wang, Anitha S Nair, Hongmei Fu, Silvia Angeletti, Silvia Spoto, Marta Fogolari, Francesco Romano, Dunja Aksentijevic, Weiwei Liu, Baiying Li, Lixin Cheng, Liwen Jiang, Juho Vuononvirta, Thanushiyan R Poobalasingam, David M Smith, Massimo Ciccozzi, Egle Solito, Federica M Marelli-Berg
ABSTRACT
Endothelial barrier (EB) breaching is a frequent event during inflammation, and it is followed by the rapid recovery of microvascular integrity. The molecular mechanisms of EB recovery are poorly understood. Triggering of MHC molecules by migrating T-cells is a minimal signal capable of inducing endothelial contraction and transient microvascular leakage. Using this model, we show that EB recovery requires a CD31 receptor-induced, robust glycolytic response sustaining junction re-annealing. Mechanistically, this response involves src-homology phosphatase activation leading to Akt-mediated nuclear exclusion of FoxO1 and concomitant β-catenin translocation to the nucleus, collectively leading to cMyc transcription. CD31 signals also sustain mitochondrial respiration, however this pathway does not contribute to junction remodeling. We further show that pathologic microvascular leakage in CD31-deficient mice can be corrected by enhancing the glycolytic flux via pharmacological Akt or AMPK activation, thus providing a molecular platform for the therapeutic control of EB response.
Sigma-Aldrich
Fibronectin bovine plasma, solution, sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Hoechst 33258 solution, 1 mg/mL in H2O, ≥98.0% (HPLC)
Sigma-Aldrich
Brij® L23, main component: tricosaethylene glycol dodecyl ether
Millipore
Protein G Sepharose, Fast Flow, recombinant, expressed in E. coli, aqueous ethanol suspension
CellCrown inserts, 6 well plate inserts with 1.0 μm polycarbonate filter, sterile
Sigma-Aldrich
2-Deoxy-D-glucose, ≥98% (GC), crystalline
Sigma-Aldrich
Phalloidin–Tetramethylrhodamine B isothiocyanate, sequence from Amanita phalloides(synthetic: peptide sequence)
Sigma-Aldrich
Evans Blue, Dye content ≥75 %
Sigma-Aldrich
Staphylococcal enterotoxin A from Staphylococcus aureus
Sigma-Aldrich
Paraformaldehyde, powder, 95%
Supelco
3-(Trimethylsilyl)propionic-2,2,3,3-d4 acid sodium salt, 98 atom % D
Sigma-Aldrich
Paraformaldehyde, meets analytical specification of DAC, 95.0-100.5%
Sigma-Aldrich
Gelatin from bovine skin, gel strength ~225 g Bloom, Type B
Sigma-Aldrich
Collagenase from Clostridium histolyticum, suitable for release of physiologically active rat hepatocytes, Type IV, 0.5-5.0 FALGPA units/mg solid, ≥125 CDU/mg solid
Corning® HTS Transwell®-24 well permeable supports, HTS Transwell-24 units w/ 0.4 μm pore polycarbonate membrane and 6.5 mm inserts, TC-treated, sterile, 2/cs
Sigma-Aldrich
Formamide, ≥99.5% (GC), BioReagent, Molecular Biology
Sigma-Aldrich
Trypsin-EDTA solution, 0.25%, sterile-filtered, BioReagent, suitable for cell culture, 2.5 g porcine trypsin and 0.2 g EDTA, 4Na per liter of Hanks′ Balanced Salt Solution with phenol red
Supelco
Histamine, analytical standard