Merck
  • Inhibition of transcription by dactinomycin reveals a new characteristic of immunogenic cell stress.
Inhibition of transcription by dactinomycin reveals a new characteristic of immunogenic cell stress.
EMBO molecular medicine (2020-04-24)
Juliette Humeau, Allan Sauvat, Giulia Cerrato, Wei Xie, Friedemann Loos, Francesca Iannantuoni, Lucillia Bezu, Sarah Lévesque, Juliette Paillet, Jonathan Pol, Marion Leduc, Laurence Zitvogel, Hugues de Thé, Oliver Kepp, Guido Kroemer
ABSTRACT
Chemotherapy still constitutes the standard of care for the treatment of most neoplastic diseases. Certain chemotherapeutics from the oncological armamentarium are able to trigger pre-mortem stress signals that lead to immunogenic cell death (ICD), thus inducing an antitumor immune response and mediating long-term tumor growth reduction. Here, we used an established model, built on artificial intelligence to identify, among a library of 50,000 compounds, anticancer agents that, based on their molecular descriptors, were predicted to induce ICD. This algorithm led us to the identification of dactinomycin (DACT, best known as actinomycin D), a highly potent cytotoxicant and ICD inducer that mediates immune-dependent anticancer effects in vivo. Since DACT is commonly used as an inhibitor of DNA to RNA transcription, we investigated whether other experimentally established or algorithm-selected, clinically employed ICD inducers would share this characteristic. As a common leitmotif, a panel of pharmacological ICD stimulators inhibited transcription and secondarily translation. These results establish the inhibition of RNA synthesis as an initial event for ICD induction.
Sigma-Aldrich
Docetaxel, purum, ≥97.0% (HPLC)
Sigma-Aldrich
IGEPAL® CO-630, average Mn 617
Sigma-Aldrich
Doxorubicin hydrochloride, 98.0-102.0% (HPLC)
Sigma-Aldrich
Actinomycin D, from Streptomyces sp., ~98% (HPLC)
Sigma-Aldrich
Formaldehyde solution, for molecular biology, 36.5-38% in H2O
Sigma-Aldrich
Tunicamycin from Streptomyces sp.
Sigma-Aldrich
Vinblastine sulfate salt, ≥97% (HPLC)
Sigma-Aldrich
Epirubicin hydrochloride, ≥90% (HPLC)
Millipore
Immobilon®-P PVDF Membrane, 1 roll, 27 cm x 3.75 m, 0.45 µm pore size, Hydrophobic PVDF Transfer Membrane for western blotting.
Sigma-Aldrich
5-Ethynyl uridine, ≥95%
Vincristine sulfate, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Mycophenolate mofetil, ≥98% (HPLC)
Sigma-Aldrich
Propidium iodide solution
Sigma-Aldrich
Paclitaxel, from semisynthetic, ≥98%
Sigma-Aldrich
Thapsigargin, ≥98% (HPLC), solid film
Sigma-Aldrich
Mitoxantrone dihydrochloride, ≥97% (HPLC)
Sigma-Aldrich
β-Lapachone, ≥98% (TLC)
Daunorubicin hydrochloride, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Crizotinib, ≥98% (HPLC)
Sigma-Aldrich
ISRIB, ≥98% (HPLC)
Cisplatin, European Pharmacopoeia (EP) Reference Standard