Merck
  • Evidence for a role for transglutaminase in Huntington's disease and the potential therapeutic implications.
Evidence for a role for transglutaminase in Huntington's disease and the potential therapeutic implications.
Neurochemistry international (2001-12-12)
Marcela V Karpuj, Mark W Becher, Lawrence Steinman
ABSTRACT
Transglutaminase (TGase) activity is increased in affected regions of brains from patients with Huntington's disease (HD). TGase activity is particularly elevated in the nucleus compared with the cytoplasm from these brains. Gamma-glutaminyl-lysyl cross-links have been detected in nuclear inclusions in HD brain, indicating that TGase may play a prominent role in the aggregation of huntingtin (htt). Attempts to ameliorate experimental disease, via inhibition of TGase in transgenic models of HD in mice, are under investigation.
Sigma-Aldrich
Microbial Transglutaminase, lyophilized powder, ≥12 units/mg protein