Merck
  • Does the lipid environment impact the open-state conductance of an engineered β-barrel protein nanopore?
Does the lipid environment impact the open-state conductance of an engineered β-barrel protein nanopore?
Biochimica et biophysica acta (2012-12-19)
Noriko Tomita, Mohammad M Mohammad, David J Niedzwiecki, Makoto Ohta, Liviu Movileanu
ABSTRACT
Using rational membrane protein design, we were recently able to obtain a β-barrel protein nanopore that was robust under an unusually broad range of experimental circumstances. This protein nanopore was based upon the native scaffold of the bacterial ferric hydroxamate uptake component A (FhuA) of Escherichia coli. In this work, we expanded the examinations of the open-state current of this engineered protein nanopore, also called FhuA ΔC/Δ4L, employing an array of lipid bilayer systems that contained charged and uncharged as well as conical and cylindrical lipids. Remarkably, systematical single-channel analysis of FhuA ΔC/Δ4L indicated that most of its biophysical features, such as the unitary conductance and the stability of the open-state current, were not altered under the conditions tested in this work. However, electrical recordings at high transmembrane potentials revealed that the presence of conical phospholipids within the bilayer catalyzes the first, stepwise current transition of the FhuA ΔC/Δ4L protein nanopore to a lower-conductance open state. This study reinforces the stability of the open-state current of the engineered FhuA ΔC/Δ4L protein nanopore under various experimental conditions, paving the way for further critical developments in biosensing and molecular biomedical diagnosis.
Product Description
Avanti
4ME 16:0 PE, Avanti Research - A Croda Brand