Merck
  • Therapeutic potential of KLF2-induced exosomal microRNAs in pulmonary hypertension.
Therapeutic potential of KLF2-induced exosomal microRNAs in pulmonary hypertension.
Nature communications (2020-03-07)
Hebah A Sindi, Giusy Russomanno, Sandro Satta, Vahitha B Abdul-Salam, Kyeong Beom Jo, Basma Qazi-Chaudhry, Alexander J Ainscough, Robert Szulcek, Harm Jan Bogaard, Claire C Morgan, Soni S Pullamsetti, Mai M Alzaydi, Christopher J Rhodes, Roberto Piva, Christina A Eichstaedt, Ekkehard Grünig, Martin R Wilkins, Beata Wojciak-Stothard
ABSTRACT
Pulmonary arterial hypertension (PAH) is a severe disorder of lung vasculature that causes right heart failure. Homoeostatic effects of flow-activated transcription factor Krüppel-like factor 2 (KLF2) are compromised in PAH. Here, we show that KLF2-induced exosomal microRNAs, miR-181a-5p and miR-324-5p act together to attenuate pulmonary vascular remodelling and that their actions are mediated by Notch4 and ETS1 and other key regulators of vascular homoeostasis. Expressions of KLF2, miR-181a-5p and miR-324-5p are reduced, while levels of their target genes are elevated in pre-clinical PAH, idiopathic PAH and heritable PAH with missense p.H288Y KLF2 mutation. Therapeutic supplementation of miR-181a-5p and miR-324-5p reduces proliferative and angiogenic responses in patient-derived cells and attenuates disease progression in PAH mice. This study shows that reduced KLF2 signalling is a common feature of human PAH and highlights the potential therapeutic role of KLF2-regulated exosomal miRNAs in PAH and other diseases associated with vascular remodelling.
Sigma-Aldrich
MISSION® esiRNA, targeting human KLF4
Sigma-Aldrich
MISSION® esiRNA, targeting human ETS1
Sigma-Aldrich
Fibronectin bovine plasma, solution, sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Triton X-100, laboratory grade
Sigma-Aldrich
p-Xylene-bis(N-pyridinium bromide), ≥95% (TLC)
Sigma-Aldrich
PKH26 Red Fluorescent Cell Linker Kit for General Cell Membrane Labeling, Distributed for Phanos Technologies
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-74, purified immunoglobulin, buffered aqueous solution
Sigma-Aldrich
Anti-Actin, α-Smooth Muscle antibody, Mouse monoclonal, clone 1A4, purified from hybridoma cell culture
BRAND® 96-well microplate, U-bottom, round bottom, non-sterile
Sigma-Aldrich
GW4869, ≥90% (NMR)
Sigma-Aldrich
Anti-Mouse IgG (whole molecule)−TRITC antibody produced in goat, IgG fraction of antiserum, buffered aqueous solution
Sigma-Aldrich
Bovine Serum Albumin, heat shock fraction, protease free, pH 7, ≥98%
Sigma-Aldrich
Bovine Serum Albumin, lyophilized powder, essentially IgG-free, low endotoxin, BioReagent, suitable for cell culture
Sigma-Aldrich
Phalloidin–Tetramethylrhodamine B isothiocyanate, sequence from Amanita phalloides(synthetic: peptide sequence)
Sigma-Aldrich
Phalloidin, Fluorescein Isothiocyanate Labeled, sequence Amanita phalloides(synthetic: peptide sequence)
Millipore
Millipore® Steriflip® Vacuum Tube Top Filter, pore size 0.45 μm, PVDF membrane, funnel capacity 50 mL, pack of 25 ea
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Notch4
Sigma-Aldrich
Anti-Rabbit IgG (whole molecule)–Peroxidase antibody produced in goat, affinity isolated antibody, buffered aqueous solution