Merck
Targeted chemotherapy overcomes drug resistance in melanoma.
Genes & development (2020-04-04)
Jingyin Yue, Roberto Vendramin, Fan Liu, Omar Lopez, Monica G Valencia, Helena Gomes Dos Santos, Gabriel Gaidosh, Felipe Beckedorff, Ezra Blumenthal, Lucia Speroni, Stephen D Nimer, Jean-Christophe Marine, Ramin Shiekhattar
ABSTRACT
The emergence of drug resistance is a major obstacle for the success of targeted therapy in melanoma. Additionally, conventional chemotherapy has not been effective as drug-resistant cells escape lethal DNA damage effects by inducing growth arrest commonly referred to as cellular dormancy. We present a therapeutic strategy termed "targeted chemotherapy" by depleting protein phosphatase 2A (PP2A) or its inhibition using a small molecule inhibitor (1,10-phenanthroline-5,6-dione [phendione]) in drug-resistant melanoma. Targeted chemotherapy induces the DNA damage response without causing DNA breaks or allowing cellular dormancy. Phendione treatment reduces tumor growth of BRAFV600E-driven melanoma patient-derived xenografts (PDX) and diminishes growth of NRASQ61R-driven melanoma, a cancer with no effective therapy. Remarkably, phendione treatment inhibits the acquisition of resistance to BRAF inhibition in BRAFV600E PDX highlighting its effectiveness in combating the advent of drug resistance.
Product Description
Deferoxamine mesylate salt, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
3-Hydroxy-1,2-dimethyl-4(1H)-pyridone, 98%
Sigma-Aldrich
N,N,N′,N′-Tetrakis(2-pyridylmethyl)ethylenediamine
Sigma-Aldrich
Deferoxamine mesylate salt, powder, ≥92.5% (TLC)
Sigma-Aldrich
Pyrene, puriss. p.a., for fluorescence, ≥99.0% (GC)
Sigma-Aldrich
Anti-phospho-ATM (Ser1981) Antibody, clone 10H11.E12, clone 10H11.E12, Upstate®, from mouse
Deferiprone, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Anti-Chk2 Antibody, clone 7, clone 7, Upstate®, from mouse
Sigma-Aldrich
1,10-Phenanthroline-5,6-dione, 97%
Sigma-Aldrich
Zinquin ethyl ester, ≥95% (HPLC), solid