Merck
  • TMEM16F phospholipid scramblase mediates trophoblast fusion and placental development.
TMEM16F phospholipid scramblase mediates trophoblast fusion and placental development.
Science advances (2020-06-05)
Yang Zhang, Trieu Le, Ryan Grabau, Zahra Mohseni, Hoejeong Kim, David R Natale, Liping Feng, Hua Pan, Huanghe Yang
ABSTRACT
Cell-cell fusion or syncytialization is fundamental to the reproduction, development, and homeostasis of multicellular organisms. In addition to various cell type-specific fusogenic proteins, cell surface externalization of phosphatidylserine (PS), a universal eat-me signal in apoptotic cells, has been observed in different cell fusion events. Nevertheless, the molecular underpinnings of PS externalization and cellular mechanisms of PS-facilitated cell-cell fusion are unclear. Here, we report that TMEM16F, a Ca2+-activated phospholipid scramblase (CaPLSase), plays an essential role in placental trophoblast fusion by translocating PS to cell surface independent of apoptosis. The placentas from the TMEM16F knockout mice exhibit deficiency in trophoblast syncytialization and placental development, which lead to perinatal lethality. We thus identified a new biological function of TMEM16F CaPLSase in trophoblast fusion and placental development. Our findings provide insight into understanding cell-cell fusion mechanism of other cell types and on mitigating pregnancy complications such as miscarriage, intrauterine growth restriction, and preeclampsia.
Sigma-Aldrich
Anti-ANO6 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Q-VD-OPh hydrate, ≥95% (HPLC)
Sigma-Aldrich
Poly-L-lysine hydrobromide, mol wt 30,000-70,000
Sigma-Aldrich
Percoll®, pH 8.5-9.5 (20 °C)
Sigma-Aldrich
TWEEN® 20, for molecular biology, viscous liquid
Sigma-Aldrich
Anti-Monocarboxylate Transporter 1 Antibody, from chicken, purified by affinity chromatography