Merck
  • Reactivity studies of 3,3-bis(trimethylsilyl)-2-methyl-1-propene in Lewis acid-catalyzed allylation reactions.
Reactivity studies of 3,3-bis(trimethylsilyl)-2-methyl-1-propene in Lewis acid-catalyzed allylation reactions.
Organic letters (2006-09-22)
David R Williams, Angel I Morales-Ramos, Catherine M Williams
ABSTRACT
Allylation reagents, which possess geminal bis-trimethylsilyl substitution, are readily prepared from E- or Z-alkenyl bromides. The reactivity of 3,3-bis(trimethylsilyl)-2-methyl-1-propene (1) is described and predominantly provides ene reactions with aldehydes to give alcohol 2 in the presence of BF3.OEt2. Alternatively, Sakurai allylation reactions of 1 are observed by using stronger Lewis acids in methylene chloride to exclusively yield E-trisubstituted alkenylsilanes 3.
Sigma-Aldrich
Chlorobis(trimethylsilyl)methane, 97%