Merck
  • Validation of a handheld β-hydroxybutyrate acid meter to identify hyperketonaemia in ewes.
Validation of a handheld β-hydroxybutyrate acid meter to identify hyperketonaemia in ewes.
PeerJ (2020-04-21)
Carolina Akiko Sato Cabral Araújo, Antonio Humberto Hamad Minervino, Rejane Santos Sousa, Francisco Leonardo Costa Oliveira, Frederico Augusto Mazzocca Lopes Rodrigues, Clara Satsuki Mori, Enrico Lippi Ortolani
ABSTRACT
The end of pregnancy is the period with the highest risk of occurrences of ketosis and pregnancy toxemia due to fat mobilization and increasing non-esterified fatty acids in the liver which are converted in ketone bodies, mainly β-hydroxybutyrate acid (BHB). This ketone body may also become elevated in the bloodstream. The present study validates the use of a handheld meter for determining the blood concentration of BHB and ascertaining the predictive value and accuracy of BHB measurements in diagnosing hyperketonaemia in ewes. A total of 19, non-pregnant, crossbred ewes were subjected to 2 h of intravenous infusion of a saturated BHB solution. Over 6 h of evaluation, 247 blood samples were obtained in 13 sampling moments. The BHB concentration was measured by an enzymatic colorimetric method in an automated biochemical analyzer (gold-standard) and by a handheld meter using an electrochemical enzyme technique. There was a high correlation between both methods (r = 0.98; P < 0.001). Considering the blood BHB concentrations range 0.8-1.6 mmol/L for moderate ketosis the handheld meter presented sensitivity and specificity of 0.98 and 0.81, respectively. For severe ketosis (BHB ≥ 1.6 mmol/L) sensitivity and specificity were 0.99 and 0.75, respectively. Thus, the handheld device can be useful for diagnoses of cases of mild or severe pregnancy toxemia at field conditions.
Sigma-Aldrich
(±)-Sodium 3-hydroxybutyrate, ≥99.0% (NT)