Merck
  • Nucleocytoplasmic Proteomic Analysis Uncovers eRF1 and Nonsense-Mediated Decay as Modifiers of ALS/FTD C9orf72 Toxicity.
Nucleocytoplasmic Proteomic Analysis Uncovers eRF1 and Nonsense-Mediated Decay as Modifiers of ALS/FTD C9orf72 Toxicity.
Neuron (2020-02-16)
Juan A Ortega, Elizabeth L Daley, Sukhleen Kour, Marisa Samani, Liana Tellez, Haley S Smith, Elizabeth A Hall, Y Taylan Esengul, Yung-Hsu Tsai, Tania F Gendron, Christopher J Donnelly, Teepu Siddique, Jeffrey N Savas, Udai B Pandey, Evangelos Kiskinis
ABSTRACT
The most common genetic cause of amyotrophic lateral sclerosis (ALS) and frontotemporal dementia (FTD) is a hexanucleotide repeat expansion in C9orf72 (C9-HRE). While RNA and dipeptide repeats produced by C9-HRE disrupt nucleocytoplasmic transport, the proteins that become redistributed remain unknown. Here, we utilized subcellular fractionation coupled with tandem mass spectrometry and identified 126 proteins, enriched for protein translation and RNA metabolism pathways, which collectively drive a shift toward a more cytosolic proteome in C9-HRE cells. Among these was eRF1, which regulates translation termination and nonsense-mediated decay (NMD). eRF1 accumulates within elaborate nuclear envelope invaginations in patient induced pluripotent stem cell (iPSC) neurons and postmortem tissue and mediates a protective shift from protein translation to NMD-dependent mRNA degradation. Overexpression of eRF1 and the NMD driver UPF1 ameliorate C9-HRE toxicity in vivo. Our findings provide a resource for proteome-wide nucleocytoplasmic alterations across neurodegeneration-associated repeat expansion mutations and highlight eRF1 and NMD as therapeutic targets in C9orf72-associated ALS and/or FTD.
Sigma-Aldrich
MISSION® esiRNA, targeting human ETF1
Sigma-Aldrich
Sodium acrylate, 97%
Sigma-Aldrich
Monoclonal Anti-Vinculin antibody produced in mouse, clone hVIN-1, ascites fluid
Sigma-Aldrich
Retinoic acid, ≥98% (HPLC), powder
Sigma-Aldrich
Doxycycline hyclate
Sigma-Aldrich
DAPT, ≥98% (HPLC), solid
Sigma-Aldrich
Y-27632 dihydrochloride, ≥98% (HPLC)
Sigma-Aldrich
MISSION® esiRNA, targeting human UPF1
Sigma-Aldrich
L-Ascorbic acid, powder, suitable for cell culture, γ-irradiated
Sigma-Aldrich
Triton X-100, laboratory grade
Sigma-Aldrich
Monoclonal Anti-α-Tubulin antibody produced in mouse, ascites fluid, clone B-5-1-2
Sigma-Aldrich
Anti-β-Tubulin III antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Anti-C9ORF72/C9RANT (poly-GR) Antibody, clone 5A2, clone 5A2, 1 mg/mL, from rat
Sigma-Aldrich
Anti-phospho-Upf1 (Ser1127) Antibody, from rabbit, purified by affinity chromatography
Sigma-Aldrich
Anti-Puromycin Antibody, clone 12D10, clone 12D10, from mouse
Roche
Anti-Digoxigenin-Fluorescein, Fab fragments, from sheep