Merck
  • Physical properties of edible emulsified films based on carboxymethyl cellulose and oleic acid.
Physical properties of edible emulsified films based on carboxymethyl cellulose and oleic acid.
International journal of biological macromolecules (2010-10-06)
Babak Ghanbarzadeh, Hadi Almasi
ABSTRACT
Glycerol and oleic acid (OA) were incorporated into carboxymethyl cellulose (CMC) films by an emulsification method. Films containing different amounts of glycerol and OA were examined for mechanical properties, water vapor permeability (WVP), and moisture uptake, optical and thermal properties. Addition of OA to the CMC films significantly improved the barrier property. However, the effect of OA on the mechanical properties was lower than glycerol. By increasing of OA content, the cloudiness of the CMC films was intensified and Hunter value (b) of the films increased (by ca. 35.8%).
Sigma-Aldrich
Carboxymethylcellulose sodium salt, ultra high viscosity, highly purified
Sigma-Aldrich
Carboxymethylcellulose sodium salt, High viscosity
Sigma-Aldrich
Carboxymethylcellulose sodium salt, Medium viscosity
Sigma-Aldrich
Carboxymethylcellulose sodium salt, medium viscosity
Sigma-Aldrich
Carboxymethylcellulose sodium salt, low viscosity