Merck
  • Characterization of Engineering Plastics Plasticized Using Supercritical CO2.
Characterization of Engineering Plastics Plasticized Using Supercritical CO2.
Polymers (2020-01-16)
Masaki Watanabe, Yoshihide Hashimoto, Tsuyoshi Kimura, Akio Kishida
ABSTRACT
The purpose of this study was to evaluate the physical and chemical properties of engineering plastics processed using supercritical CO2. First, we prepared disk-shaped test pieces via a general molding process, which were plasticized using supercritical CO2 at temperatures lower than the glass-transition points of engineering plastics. Amorphous polymers were plasticized, and their molecular weight remained nearly unchanged after treatment with supercritical CO2. The mechanical strength significantly decreased despite the unchanged molecular weight. The surface roughness and contact angle increased slightly, and electrical properties such as the rate of charging decreased significantly. These results suggest that supercritical CO2 could be used for a new molding process performed at lower temperatures than those used in general molding processes, according to the required properties.
Sigma-Aldrich
Poly(1,4-butylene terephthalate), average Mv ~38,000, pellets
Pricing and availability is not currently available.
Sigma-Aldrich
Poly(ethylene terephthalate), granular
Pricing and availability is not currently available.
Sigma-Aldrich
Polysulfone, average Mn ~22,000 by MO, beads
Pricing and availability is not currently available.
Sigma-Aldrich
Polyetherimide, melt index 9 g/10 min (337 °C/6.6kg)
Pricing and availability is not currently available.