Merck
  • Determination of 5alpha-androst-16-en-3alpha-ol in truffle fermentation broth by solid-phase extraction coupled with gas chromatography-flame ionization detector/electron impact mass spectrometry.
Determination of 5alpha-androst-16-en-3alpha-ol in truffle fermentation broth by solid-phase extraction coupled with gas chromatography-flame ionization detector/electron impact mass spectrometry.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences (2008-07-01)
Guan Wang, Yuan-Yuan Li, Dong-Sheng Li, Ya-Jie Tang
ABSTRACT
A novel method using solid-phase extraction coupled with gas chromatography and flame ionization detector (FID)/electron impact mass spectrometry (EIMS) was developed for the determination of 5alpha-androst-16-en-3alpha-ol (androstenol), a steroidal compound belonging to the group of musk odorous 16-androstenes, in truffle fermentation broth. Comparison studies between FID and EIMS indicated two detectors gave similar quantitative results. The highest androstenol concentration of 123.5 ng/mL was detected in Tuber indicum fermentation broth, while no androstenol was found in Tuber aestivum fermentation broth. For the first time, this work confirmed the existence of androstenol in the truffle fermentation broth, which suggested truffle fermentation is a promising alternative for androstenol production on a large scale.
Sigma-Aldrich
5α-Androst-16-en-3α-ol
Pricing and availability is not currently available.