Merck
  • Restraining Lysosomal Activity Preserves Hematopoietic Stem Cell Quiescence and Potency.
Restraining Lysosomal Activity Preserves Hematopoietic Stem Cell Quiescence and Potency.
Cell stem cell (2020-02-29)
Raymond Liang, Tasleem Arif, Svetlana Kalmykova, Artem Kasianov, Miao Lin, Vijay Menon, Jiajing Qiu, Jeffrey M Bernitz, Kateri Moore, Fangming Lin, Deanna L Benson, Nikolaos Tzavaras, Milind Mahajan, Dmitri Papatsenko, Saghi Ghaffari
ABSTRACT
Quiescence is a fundamental property that maintains hematopoietic stem cell (HSC) potency throughout life. Quiescent HSCs are thought to rely on glycolysis for their energy, but the overall metabolic properties of HSCs remain elusive. Using combined approaches, including single-cell RNA sequencing (RNA-seq), we show that mitochondrial membrane potential (MMP) distinguishes quiescent from cycling-primed HSCs. We found that primed, but not quiescent, HSCs relied readily on glycolysis. Notably, in vivo inhibition of glycolysis enhanced the competitive repopulation ability of primed HSCs. We further show that HSC quiescence is maintained by an abundance of large lysosomes. Repression of lysosomal activation in HSCs led to further enlargement of lysosomes while suppressing glucose uptake. This also induced increased lysosomal sequestration of mitochondria and enhanced the competitive repopulation ability of primed HSCs by over 90-fold in vivo. These findings show that restraining lysosomal activity preserves HSC quiescence and potency and may be therapeutically relevant.
Sigma-Aldrich
Tetramethylrhodamine ethyl ester perchlorate, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
Methyl pyruvate, 90%, technical grade
Sigma-Aldrich
α-Cyano-4-hydroxycinnamic acid, ≥98% (TLC), powder
Sigma-Aldrich
Carbonyl cyanide 3-chlorophenylhydrazone, ≥97% (TLC), powder
Sigma-Aldrich
Pyronin Y, suitable for microscopy
Sigma-Aldrich
STF-31, ≥98% (HPLC)
Sigma-Aldrich
2-Deoxy-D-glucose, ≥98% (GC), crystalline
Sigma-Aldrich
Doxycycline hyclate
Sigma-Aldrich
Dimethyl 2-oxoglutarate, 96%
Sigma-Aldrich
Chloroquine diphosphate salt, powder or crystals, 98.5-101.0% (EP)
Sigma-Aldrich
Leupeptin, microbial, ≥90% (HPLC)
Sigma-Aldrich
Propidium iodide, ≥94.0% (HPLC)