Merck
  • AIM2 promotes the development of non-small cell lung cancer by modulating mitochondrial dynamics.
AIM2 promotes the development of non-small cell lung cancer by modulating mitochondrial dynamics.
Oncogene (2020-02-02)
Miao Qi, Dan Dai, Jin Liu, Zhongqi Li, Panpan Liang, Yue Wang, Lu Cheng, Yihong Zhan, Zhifeng An, Yaoyao Song, Yana Yang, Xiaohui Yan, Hui Xiao, Huanjie Shao
ABSTRACT
Mitochondrial fusion and fission dynamics fine-tune cellular calcium homeostasis, ATP production capacity and ROS production and play important roles in cell proliferation and migration. Dysregulated mitochondrial dynamics is closely related to tumor development, but the mechanism of mitochondrial dynamics dysregulation and its role in the development of lung cancer remains unclear. Here, we demonstrate that the DNA sensor protein absent in melanoma 2 (AIM2) is highly expressed in non-small cell lung cancer (NSCLC) cells and that high AIM2 expression is associated with poor prognosis in patients with NSCLC. High expression of AIM2 contributes to tumor cell growth and proliferation independent of inflammasome activation in vitro and in vivo. Further studies have shown that AIM2 colocalizes with mitochondria in NSCLC cells and that AIM2 knockdown leads to enhanced mitochondrial fusion and decreased cell proliferation. Mechanistic studies have shown that AIM2 downregulation promotes MFN2 upregulation, thereby enhancing mitochondrial fusion. Moreover, we found that mitochondrial fusion driven by AIM2 knockdown leads to a decrease of cellular reactive oxygen species (ROS) production, which further causes inactivation of the MAPK/ERK signaling pathway. Together, we discovered a novel function of AIM2 in promoting NSCLC development by regulating mitochondrial dynamics and revealed its underlying mechanism. Our work provides new intervention targets for the treatment of NSCLC.
Sigma-Aldrich
Hydrogen peroxide solution, contains ~200 ppm acetanilide as stabilizer, 3 wt. % in H2O
Sigma-Aldrich
MISSION® esiRNA, targeting human AIM2
Sigma-Aldrich
Amyloid Protein Non-Aβ Component, ≥80% (HPLC)
Sigma-Aldrich
N-Acetyl-L-cysteine, Sigma Grade, ≥99% (TLC), powder