Merck
  • Non-anticoagulant heparin increases endothelial nitric oxide synthase activity: role of inhibitory guanine nucleotide proteins.
Non-anticoagulant heparin increases endothelial nitric oxide synthase activity: role of inhibitory guanine nucleotide proteins.
Journal of molecular and cellular cardiology (1999-02-17)
P C Kouretas, R L Hannan, N K Kapur, R Hendrickson, E M Redmond, A K Myers, Y D Kim, P A Cahill, J V Sitzmann
ABSTRACT
Heparin, which is widely used clinically, has recently been shown to have specific properties affecting the vascular endothelium. We hypothesized that heparin stimulates endothelial nitric oxide synthase (eNOS) activity by a mechanism independent of its anticoagulant properties and dependent on an inhibitory guanine nucleotide regulatory protein (Gi). We determined the effect of both heparin and N-acetyl heparin (Non-Hep), a heparin derivative without anticoagulant properties, on eNOS activity in cultured bovine aortic endothelial cells and on endothelium-dependent relaxation in isolated vascular rings. The eNOS activity was determined by measuring both citrulline and nitric oxide (NO) metabolite formation. Heparin and Non-Hep dose-dependently increased basal eNOS activity (ED50 1.0 microgram/ml or 0.15 U/ml), an effect that was significantly inhibited by pertussis toxin (100 ng/ml), a Gi-protein inhibitor. Agonist-stimulated (acetylcholine, 10 microM) eNOS activity was potentiated following pre-treatment with both heparin and Non-Hep and reversed by pertussis toxin. Heparin and Non-Hep induced a dose-dependent relaxation in preconstricted thoracic aortic rings, an effect that was significantly inhibited by pertussis toxin, endothelial inactivation (following treatment with sodium deoxycholate) and NG-nitro-L-arginine-methyl ester (L-NAME). We conclude that heparin and non-anticoagulant heparin induce endothelium-dependent relaxation following activation of eNOS by a mechanism involving a Gi-protein. Administration of heparin derivatives without anticoagulant properties may have therapeutic implications for the preservation of eNOS in conditions characterized by endothelial dysfunction.
Sigma-Aldrich
N-Acetylheparin sodium salt, ~90%