Merck
A High-Throughput Screening Identifies MICU1 Targeting Compounds.
Cell reports (2020-02-23)
Giulia Di Marco, Francesca Vallese, Benjamin Jourde, Christian Bergsdorf, Mattia Sturlese, Agnese De Mario, Valerie Techer-Etienne, Dorothea Haasen, Berndt Oberhauser, Simone Schleeger, Giulia Minetti, Stefano Moro, Rosario Rizzuto, Diego De Stefani, Mara Fornaro, Cristina Mammucari
ABSTRACT
Mitochondrial Ca2+ uptake depends on the mitochondrial calcium uniporter (MCU) complex, a highly selective channel of the inner mitochondrial membrane (IMM). Here, we screen a library of 44,000 non-proprietary compounds for their ability to modulate mitochondrial Ca2+ uptake. Two of them, named MCU-i4 and MCU-i11, are confirmed to reliably decrease mitochondrial Ca2+ influx. Docking simulations reveal that these molecules directly bind a specific cleft in MICU1, a key element of the MCU complex that controls channel gating. Accordingly, in MICU1-silenced or deleted cells, the inhibitory effect of the two compounds is lost. Moreover, MCU-i4 and MCU-i11 fail to inhibit mitochondrial Ca2+ uptake in cells expressing a MICU1 mutated in the critical amino acids that forge the predicted binding cleft. Finally, these compounds are tested ex vivo, revealing a primary role for mitochondrial Ca2+ uptake in muscle growth. Overall, MCU-i4 and MCU-i11 represent leading molecules for the development of MICU1-targeting drugs.
Sigma-Aldrich
Anti-MICU2 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Amyloglucosidase from Aspergillus niger, lyophilized, powder, ~70 U/mg
Sigma-Aldrich
Tetramethylrhodamine methyl ester perchlorate, ≥95%
Sigma-Aldrich
Histamine dihydrochloride, ≥99% (TLC), powder