Merck
  • Quantification, source apportionment and risk assessment of polycyclic aromatic hydrocarbons in sediments from Mokolo and Blood Rivers in Limpopo Province, South Africa.
Quantification, source apportionment and risk assessment of polycyclic aromatic hydrocarbons in sediments from Mokolo and Blood Rivers in Limpopo Province, South Africa.
Journal of environmental science and health. Part A, Toxic/hazardous substances & environmental engineering (2019-09-19)
Tumelo M Mogashane, Munyaradzi Mujuru, Robert I McCrindle, Abayneh A Ambushe
ABSTRACT
The presence of polycyclic aromatic hydrocarbons (PAHs) in the environment is of major concern since some of these compounds are highly persistent, toxic (causing cancer) and wide spread pollutants. The objective of this study was to evaluate the levels of PAHs in sediment samples collected from Blood and Mokolo Rivers in Limpopo Province, South Africa. The PAHs in sediments were extracted using optimized microwave-assisted extraction (MAE) method. The quantification of sixteen (16) PAHs in sediments was done by gas chromatography-flame ionization detector (GC-FID). The levels of PAHs recorded in sediment samples from Blood River ranged between 0.015 and 3.25 mg kg-1. The concentrations of PAHs obtained in sediments from Mokolo River (0.047 to 52.7 mg kg-1) were higher than those recorded in sediments from Blood River. The PAHs ratios indicated that both pyrogenic and petrogenic could be the sources of these compounds in both rivers. Toxic equivalency factors (TEFs) and benzo(a)pyrene equivalent (BaPE) were used to estimate the potential human health risk of PAHs in quantitative terms. The assessment of ecotoxicological risk indicated that the sediment samples collected from Mokolo River are at high toxicity risk while sediments from Blood River are at low sediment toxicity risk.
Product Description
Supelco
PAH - Sediment 1, certified reference material, pkg of 50 g