Merck
  • Spatially and temporally defined lysosomal leakage facilitates mitotic chromosome segregation.
Spatially and temporally defined lysosomal leakage facilitates mitotic chromosome segregation.
Nature communications (2020-01-15)
Saara Hämälistö, Jonathan Lucien Stahl, Elena Favaro, Qing Yang, Bin Liu, Line Christoffersen, Ben Loos, Claudia Guasch Boldú, Johanna A Joyce, Thomas Reinheckel, Marin Barisic, Marja Jäättelä
ABSTRACT
Lysosomes are membrane-surrounded cytoplasmic organelles filled with a powerful cocktail of hydrolases. Besides degrading cellular constituents inside the lysosomal lumen, lysosomal hydrolases promote tissue remodeling when delivered to the extracellular space and cell death when released to the cytosol. Here, we show that spatially and temporally controlled lysosomal leakage contributes to the accurate chromosome segregation in normal mammalian cell division. One or more chromatin-proximal lysosomes leak in the majority of prometaphases, after which active cathepsin B (CTSB) localizes to the metaphase chromatin and cleaves a small subset of histone H3. Stabilization of lysosomal membranes or inhibition of CTSB activity during mitotic entry results in a significant increase in telomere-related chromosome segregation defects, whereas cells and tissues lacking CTSB and cells expressing CTSB-resistant histone H3 accumulate micronuclei and other nuclear defects. These data suggest that lysosomal leakage and chromatin-associated CTSB contribute to proper chromosome segregation and maintenance of genomic integrity.
Sigma-Aldrich
L-Buthionine-sulfoximine, ≥97% (TLC)
Sigma-Aldrich
DL-Dithiothreitol, for molecular biology, ≥98% (HPLC), ≥99% (titration)
Sigma-Aldrich
Filipin III from Streptomyces filipinensis, ≥85% (HPLC)
Sigma-Aldrich
Duolink® In Situ PLA® Probe Anti-Mouse PLUS
Sigma-Aldrich
Duolink® In Situ PLA® Probe Anti-Rabbit MINUS
Sigma-Aldrich
Dipeptidyl Peptidase III human, recombinant, expressed in Sf9 cells
Sigma-Aldrich
Methyl cellulose, viscosity: 15 cP
Sigma-Aldrich
MISSION® esiRNA, targeting human CTSB
Sigma-Aldrich
3-Hydroxy-1,2-dimethyl-4(1H)-pyridone, 98%
Sigma-Aldrich
MISSION® esiRNA, targeting EGFP
Sigma-Aldrich
CRISPR/Cas9 Products and Services, Design and order CRISPR gRNA, Cas9, screening libraries, controls and companion products. Formats include plant, lentivirus, IVT-RNA, plasmid, synthetic, and protein.
Sigma-Aldrich
Estrone, ≥99%
Sigma-Aldrich
(±)-α-Tocopherol, synthetic, ≥96% (HPLC)
Sigma-Aldrich
Cholesterol, powder, BioReagent, suitable for cell culture, ≥99%
Sigma-Aldrich
MISSION® esiRNA, targeting human TERF2
Sigma-Aldrich
DL-Cysteine, technical grade
Sigma-Aldrich
Phe-Arg β-naphthylamide dihydrochloride, cathepsin substrate
Sigma-Aldrich
Triton X-100, laboratory grade
Sigma-Aldrich
Duolink® In Situ Detection Reagents Red
Supelco
Simvastatin, analytical standard
Sigma-Aldrich
Cathepsin L Inhibitor IV, The Cathepsin L Inhibitor IV controls the biological activity of Cathepsin L. This small molecule/inhibitor is primarily used for Protease Inhibitors applications.
Sigma-Aldrich
Cdk1 Inhibitor IV, RO-3306, InSolution, ≥95%
Sigma-Aldrich
ALLN, Cell-permeable inhibitor of calpain I (Ki = 190 nM), calpain II (Ki = 220 nM), cathepsin B (Ki = 150 nM), and cathepsin L (Ki = 500 pM).
Sigma-Aldrich
CA-074 Me, CA-074 Me, CAS 147859-80-1, is a cell-permeable analog of CA-074 that acts as an irreversible inhibitor of intracellular cathepsin B.
Sigma-Aldrich
bisBenzimide H 33342 trihydrochloride, ≥98% (HPLC and TLC)
Sigma-Aldrich
Triton X-100, BioXtra