Merck
  • Oviduct fluid extracellular vesicles regulate polyspermy during porcine in vitro fertilisation.
Oviduct fluid extracellular vesicles regulate polyspermy during porcine in vitro fertilisation.
Reproduction, fertility, and development (2019-11-30)
A S Alcântara-Neto, M Fernandez-Rufete, E Corbin, G Tsikis, R Uzbekov, A S Garanina, P Coy, C Almiñana, P Mermillod
ABSTRACT
High polyspermy is one of the major limitations of porcine invitro fertilisation (IVF). The addition of oviductal fluid (OF) during IVF reduces polyspermy without decreasing the fertilisation rate. Because extracellular vesicles (EVs) have been described as important OF components, the aim of this study was to evaluate the effect of porcine oviductal EVs (poEVs) on IVF efficiency compared with porcine OF (fresh and lyophilised). OF was collected from abattoir oviducts by phosphate-buffered saline flush, and poEVs were isolated by serial ultracentrifugation. Four IVF treatments were conducted: poEVs (0.2mgmL-1), OF (10%), lyophilized and reconstituted pure OF (LOF; 1%) and IVF without supplementation (control). Penetration, monospermy and IVF efficiency were evaluated. Transmission electron microscopy showed an EVs population primarily composed of exosomes (83%; 30-150nm). Supplementation with poEVs during IVF increased monospermy compared with control (44% vs 17%) while maintaining an acceptable penetration rate (61% vs 78% respectively) in a similar way to OF and LOF. Western blotting revealed poEVs proteins involved in early reproductive events, including zona pellucida hardening. In conclusion, our finding show that poEVs are key components of porcine OF and may play roles in porcine fertilisation and polyspermy regulation, suggesting that supplementation with poEVs is a reliable strategy to decrease porcine polyspermy and improve invitro embryo production outcomes.
Sigma-Aldrich
Cysteamine, ~95%
Sigma-Aldrich
Gentamicin solution, 10 mg/mL in deionized water, liquid, 0.1 μm filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Medium 199, HEPES Modification, with Earle′s salts, 25 mM HEPES and sodium bicarbonate, without L-glutamine, liquid, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Medium 199, With Earle′s salts, L-glutamine and sodium bicarbonate, liquid, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Lacmoid
Sigma-Aldrich
Epidermal Growth Factor from murine submaxillary gland, EGF, suitable for cell culture