Merck
  • Quality evaluation of table grapes during storage by using 1H NMR, LC-HRMS, MS-eNose and multivariate statistical analysis.
Quality evaluation of table grapes during storage by using 1H NMR, LC-HRMS, MS-eNose and multivariate statistical analysis.
Food chemistry (2020-02-02)
Valentina Innamorato, Francesco Longobardi, Salvatore Cervellieri, Maria Cefola, Bernardo Pace, Imperatrice Capotorto, Vito Gallo, Antonino Rizzuti, Antonio F Logrieco, Vincenzo Lippolis
ABSTRACT
Three non-targeted methods, i.e. 1H NMR, LC-HRMS, and HS-SPME/MS-eNose, combined with chemometrics, were used to classify two table grape cultivars (Italia and Victoria) based on five quality levels (5, 4, 3, 2, 1). Grapes at marketable quality levels (5, 4, 3) were also discriminated from non-marketable quality levels (2 and 1). PCA-LDA and PLS-DA were applied, and results showed that, the MS-eNose provided the best results. Specifically, with the Italia table grapes, mean prediction abilities ranging from 87% to 88% and from 98% to 99% were obtained for discrimination amongst the five quality levels and of marketability/non-marketability, respectively. For the cultivar Victoria, mean predictive abilities higher than 99% were achieved for both classifications. Good models were also obtained for both cultivars using NMR and HRMS data, but only for classification by marketability. Satisfying models were further validated by MCCV. Finally, the compounds that contributed the most to the discriminations were identified.
Sigma-Aldrich
2-Methylpentanal, 98%
Sigma-Aldrich
Poly(dimethylsiloxane), viscosity 1.0 cSt (25 °C)