Merck
  • Experimental and theoretical insight into hydroxyl and sulfate radicals-mediated degradation of carbamazepine.
Experimental and theoretical insight into hydroxyl and sulfate radicals-mediated degradation of carbamazepine.
Environmental pollution (Barking, Essex : 1987) (2019-11-26)
Ruiyang Xiao, Junye Ma, Zonghao Luo, Weizhi Zeng, Zongsu Wei, Richard Spinney, Wei-Ping Hu, Dionysios D Dionysiou
ABSTRACT
Carbamazepine (CBZ), a widely detected pharmaceutical in wastewaters, cannot currently be treated by conventional activated sludge technologies, as it is highly resistant to biodegradation. In this study, the degradation kinetics and reaction mechanisms of CBZ by hydroxyl radical (OH) and sulfate radical ()-based advanced oxidation processes (AOPs) were investigated with a combined experimental/theoretical approach. We first measured the UV absorption spectrum of CBZ and compared it to the theoretical spectrum. The agreement of two spectra reveals an extended π-conjugation system on CBZ molecular structure. The second-order rate constants of OH and with CBZ, measured by competition kinetics method, were (4.63 ± 0.01) × 109 M-1 s-1 and (8.27 ± 0.01) × 108 M-1 s-1, respectively at pH 3. The energetics of the initial steps of CBZ reaction with OH and were also calculated by density functional theory (DFT) at SMD/M05-2X/6-311++G**//M05-2X/6-31 + G**level. Our results reveal that radical addition is the dominant pathway for both OH and . Further, compared to the positive ΔGR0 value for the single electron transfer (SET) reaction pathway between CBZ and OH, the ΔGR0 value for SET reaction between CBZ and is negative, showing that this reaction route is thermodynamically favorable. Our results demonstrated the remarkable advantages of AOPs for the removal of refractory organic contaminants during wastewater treatment processes. The elucidation of the pathways for the reaction of OH and with CBZ are beneficial to predict byproducts formation and assess associated ecotoxicity, providing an evaluation mean for the feasibility of AOPs application.
Sigma-Aldrich
4-Chlorobenzoic acid, 99%